Red Lion 1-16 DIN Single Dual or Triple Preset Counters
The Model C48 Counter is available as a Standard Counter or a Batch Counter. The Standard Counter is available with single or dual presets. The Batch Counter has a main process counter with dual presets and a secondary counter with a single preset. The secondary counter can be selected to function as a batch or a total counter. The C48C features a 7 segment, 2 line by 6 digit reflective or backlit LCD display. For the backlit versions, the main display line is red and shows the count value or the Batch/Total value when preset 3 or output 3 is viewed in the secondary display. The smaller secondary display line is green and can be used to view the prescaler value, preset values, output time values or Batch/Total count values (Batch model).
$222.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DIN Size Input Type Max DC Voltage VDC (VDC)
Availability Pricing
Red Lion C48CS003 1/16-DIN 1-Preset Counter, Reflective, NPN EW-68483-78 Mfr # C48CS003
COMPARE
1/16"
$222.00USD / Each
Red Lion C48CS013 1/16-DIN 1-Preset Counter, Backlit, NPN EW-68483-79 Mfr # C48CS013
COMPARE
1/16"
$222.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat