This product is now discontinued, View Related Products
Red Lion 1/8-DIN Analog Input Panel Meters
Field-upgradeable option cards offer customized solutions for any process needs
  • Temperature, process, voltage, and current inputs
  • Variable intensity display
  • 16-point scaling for non-linear processes
  • Programmable function keys and user inputs
  • 9-digit totalizer with batching
Red Lion panel meters offer many features and performance capabilities to suit a wide range of industrial applications. The meters have been specifically designed for harsh industrial environments, with NEMA 4X/IP65-rated sealed bezel and extensive testing of noise effects to CE requirements, providing a tough yet reliable application solution. Meters come with either a sunlight-readable red or a standard green LED display. The intensity of display can be adjusted from dark room applications up to outdoor use, making them ideal for viewing in bright light applications. The meters provide a Max and Min reading memory with programmable capture time. The capture time is used to prevent detection of false max or min readings which may occur during start-up or unusual process events. The signa
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Power VAC (VAC) Description
Availability Pricing
Red Lion PAXT0100 Panel Meter with Thermocouple and RTD Inputs; Green LCD EW-65590-04 Mfr # PAXT0100
COMPARE
0.56\" LED 250 Panel Meter with Thermocouple and RTD Inputs; Green LCD
Call For Price
Red Lion PAXT0110 Panel Meter with Thermocouple and RTD Inputs; DC Power; Green LCD EW-65590-06 Mfr # PAXT0110
COMPARE
0.56\" LED 250 Panel Meter with Thermocouple and RTD Inputs; DC Power; Green LCD
Call For Price
Red Lion PAXP0100 Panel Meter with mA and VDC Inputs; Green LCD EW-65590-12 Mfr # PAXP0100
COMPARE
0.56\" LED 250 Panel Meter with mA and VDC Inputs; Green LCD
Call For Price
Red Lion PAXP0110 Panel Meter with mA and VDC Inputs; DC Power; Green LCD EW-65590-14 Mfr # PAXP0110
COMPARE
0.56\" LED 250 Panel Meter with mA and VDC Inputs; DC Power; Green LCD
Call For Price
Red Lion PAXH0100 Panel Meter with RMS Volt and Current Inputs; Green LCD EW-65590-18 Mfr # PAXH0100
COMPARE
0.56\" LED 250 Panel Meter with RMS Volt and Current Inputs; Green LCD
Call For Price
Red Lion PAXCDS10 Dual Relay Option Card For Pax Series Panel Meters EW-65590-87 Mfr # PAXCDS10
COMPARE
Dual Relay Option Card For Pax Series Panel Meters