This product is now discontinued, View Related Products
Red Lion Base Mount Kits for CUB4 or CUB5 Meters
Provides mounting alternatives including DIN rail option
  • Compatible with CUB4, CUB4, or DT8 series meters
These adapter kits are designed to offer an alternative mounting option for Red Lion’s CUB4 indicator/meters and CUB5 meters.
Easy to assemble and wire! The wires can either be brought through the panel on which the meter is mounted, or through the holes in the enclosure itself. Grommets are provided to insert in the holes, along with four nuts and bolts for mounting.
BMK6 open-base mount accommodates only the meter. BMK7 closed-base mounts are large enough to accommodate both the meter and a micro-line power supply (MLPS). Constructed of rugged steel with textured black finish.
BMK11 can be used to mount either a CUB5 meter or an MLPS in various applications — it provides both base mounting and DIN rail mounting. The adapter and DIN rail clips are constructed of molded polycarbonate plastic.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Red Lion BMK40000 BASE MOUNT KITAPOLLO/GEMINI EW-68483-63 Mfr # BMK40000
COMPARE
BASE MOUNT KITAPOLLO/GEMINI
Call For Price
Red Lion BMK60000 Open Base Mount Kit for CUB4 and CUB5 EW-68516-19 Mfr # BMK60000
COMPARE
Open Base Mount Kit for CUB4 and CUB5
Call For Price
Red Lion BMK70000 Closed Base Mount Kit for CUB4 EW-68516-20 Mfr # BMK70000
COMPARE
Closed Base Mount Kit for CUB4
Call For Price
Red Lion BMK7A000 Closed Base Mount Kit for CUB5 EW-68516-21 Mfr # BMK7A000
COMPARE
Closed Base Mount Kit for CUB5
Call For Price
Red Lion BMK11000 DIN Rail Base Mount Kit EW-68516-22 Mfr # BMK11000
COMPARE
DIN Rail Base Mount Kit