This product is now discontinued, View Related Products
Red Lion Large Display Counter -Rate Meter
The large displays can be configured as a single or dual counter with rate indication, scaling, and a relay output in the 6-digit version. The 4-digit version is a single counter only(no scaling or output). The 4 and 6 digit display are available in either 2.25" or 4" high red LED digits. All versions are constructed of a NEMA 4 enclosurein light weight aluminum.
The 6-digit unit has two separate inputs which provide different functions depending on which of the eight operating modes are selected. The operating modes include bidirectional and quadrature counting as well as a count/rate indicator mode. When programmed as a dual counter, each counter has a separate scale factor and decimal points. In the count/rate indicator mode, each have their own scaling and decimal point read-outs in different engineering units.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
DIN Size Input Type Max DC Voltage VDC (VDC)
Availability Pricing
Red Lion LD200400 4DIGIT Lrg Display Counter 2.25in.High 4-Digit Red Led EW-68483-15 Mfr # LD200400
COMPARE
Call For Price
Red Lion LD2006P0 6DIGIT Lrg Display Counter/Rate Meter 2.25in.High 6-Digit Red Led EW-68483-16 Mfr # LD2006P0
COMPARE
Call For Price
Red Lion LD400400 4DIGIT Lrg Display Counter 4in.High 4-Digit Red Led EW-68483-23 Mfr # LD400400
COMPARE
Call For Price
Red Lion LD4006PO 6DIGIT Lrg Display Counter/Rate Meter 4in.High 6-Digit Red Led EW-68483-24 Mfr # LD4006PO