This product is now discontinued, View Related Products

Red Lion Magnetic Pick-Up Sensor Accessories

  • Logic magnetic pickup
  • Detects steel sensing gears or other ferrous targets
  • Built-in amplifier for low-speed operation
  • Stainless steel case
  • Epoxy encapsulated for oil, dirt, & moisture resistance
  • 9 to 17 VDC powered
Logic Magnetic Pickups (LMPs) provide sensing
sensitivities several orders of magnitude greater than
standard magnetic pickups when detecting moving
ferrous targets. By locating a high gain pre-amplifier
& signal processing circuit “inside” the mag pickup
housing, larger airgaps and slower target surface
speeds can be achieved thereby greatly expanding
application versatility as compared with conventional
mag pickups. In addition, these units have excellent
noise immunity and a frequency response to 10 KHz.
Two output types are available. The NPN Open
Collector Transistor Output unit provides a negative
going current sinking output with the approach of a
ferrous target and is current limited to 40 mA. The
Transistor Emitter-Follower Output unit provides
positive going 5 V pulses with the approach of a ferrous
target and can source 20 mA of load current. The
Open Collector units are available with either an integrally
potted 10, 25 or 50 foot 3-wire shielded cable
with neoprene strain relief boot or a polarized 3-pin
5⁄8''-24 threaded connector for quick change versatility.
The Emitter-Follower output unit is available in
the 3-pin connector version only.
Call For Price

8 variations of this product are available.

See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Red Lion 970375 20/60 Sensing Gear EW-68483-11 Mfr # 0970375
COMPARE
20/60 Sensing Gear
Call For Price
Red Lion CCA3PC00 Magnetic Pick-Up Sensor Cable, 10-Ft EW-68483-20 Mfr # CCA3PC00
COMPARE
Magnetic Pick-Up Sensor Cable, 10-Ft
Call For Price
Red Lion CCA3PC25 Magnetic Pick-Up Sensor Cable, 25-Ft EW-68483-21 Mfr # CCA3PC25
COMPARE
Magnetic Pick-Up Sensor Cable, 25-Ft
Call For Price
Red Lion CCA3PC50 Magnetic Pick-Up Sensor Cable, 50-Ft EW-68483-22 Mfr # CCA3PC50
COMPARE
Magnetic Pick-Up Sensor Cable, 50-Ft
Call For Price
Red Lion 5400100 Magnetic Pick-Up Sensor Block Mount EW-68483-35 Mfr # 5400100
COMPARE
Magnetic Pick-Up Sensor Block Mount
Call For Price
Red Lion 5403701 Magnetic Pick-Up Sensor Steel Plug Mount EW-68483-36 Mfr # 5403701
COMPARE
Magnetic Pick-Up Sensor Steel Plug Mount
Call For Price
Red Lion 5403702 Magnetic Pick-Up Sensor Stainless Steel Plug Mount EW-68483-37 Mfr # 5403702
COMPARE
Magnetic Pick-Up Sensor Stainless Steel Plug Mount
Call For Price
Red Lion MP25TA00 Magnet Pickup, 1/4in Threaded (1/4" threaded) EW-68485-59 Mfr # MP25TA00
COMPARE
Magnet Pickup, 1/4in Threaded (1/4\" threaded)
Call For Price
Showing 1- 8 of 8

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat