This product is now discontinued, View Related Products
Red Lion Magnetic Pick-Up Sensors
Logic Magnetic Pickups (LMPs) provide sensing sensitivities several orders of magnitude greater than standard magnetic pickups when detecting moving ferrous targets. By locating a high gain pre-amplifier & signal processing circuit "inside"the mag pickup housing, larger airgaps and slower target surface speeds can be achieved thereby greatly expanding application versatility as compared with conventional mag pickups. In addition, these units have excellent noise immunity and a frequency response to 10 KHz. The stainless steel sensing pole is mounted flush to the plastic sensing face allowing greater ease of setting airgaps and eliminating snagging the pole with a moving target.
Two output types are available (see ordering information). The NPN Open Collector Transistor Output unit provides a negative going current sinking output with the approach of a ferrous target and is current limited to 40 mA. The Transistor Emitter-Follower Output unit provides positive going 5 V pulses with the approach of a ferrous target and can source 20 mA of load current. The Open Collector units are available with either an integrally potted 10, 25 or 50 foot 3-wire shielded cable with neoprene strain relief boot or a polarized 3-pin 5/8"-24 threaded connector for quick change versatility. The Emitter-Follower output unit is available in the 3-pin connector version only.
FEATURES
  • Logic magnetic pickup
  • detects steel sensing gears or other ferrous targets
  • Built-in amplifier for low-speed operation
  • Stainless steel case
  • Epoxy encapsulated for oil, dirt and moisture resistance
  • 9 to 30 VDC powered
  • Call For Price
    5 variations of this product are available.
    See All Product Options Below
    5 PRODUCT OPTIONS
    Compareitem specifications
    Max Operating Temperature (° F) Max Supply Voltage (VDC) Description
    Availability Pricing
    Red Lion MPC0000 Magnetic Pickup Logic Npn O.C.10ft. Cable EW-68485-79 Mfr # LMPC0000
    COMPARE
    140 30 Magnetic Pickup Logic Npn O.C.10ft. Cable
    Call For Price
    Red Lion MPC0025 Magnetic Pickup Logic Npn O.C.25ft. Cable EW-68485-80 Mfr # LMPC0025
    COMPARE
    140 30 Magnetic Pickup Logic Npn O.C.25ft. Cable
    Call For Price
    Red Lion MPC0050 Magnetic Pickup Logic Npn O.C.50ft. Cable EW-68485-81 Mfr # LMPC0050
    COMPARE
    140 30 Magnetic Pickup Logic Npn O.C.50ft. Cable
    Call For Price
    Red Lion MPCC000 Magnetic Pickup Logic Npn O.C. 3pin Connector EW-68485-82 Mfr # LMPCC000
    COMPARE
    140 30 Magnetic Pickup Logic Npn O.C. 3pin Connector
    Call For Price
    Red Lion MPEC000 Magnetic Pickup Logic Emitter Follower 3-Pin Connector EW-68485-83 Mfr # LMPEC000
    COMPARE
    140 30 Magnetic Pickup Logic Emitter Follower 3-Pin Connector