Red Lion Miniature DC Voltage or Current Panel Meters
The CUB4 Volt and Current Meters are designed and manufactured using the latest technology for a high quality, compact, affordable instrument for use in industrial environments. Each unit has a 3 1/2-digit LCD display with 0.6 inch (15.2 mm) high digits and a DIP switch selectable decimal point. The displays are available in positive image reflective (black digits, reflective background) or negative image transmissive (illuminated digits, dark background) with red or yellow/green backlighting.
The units are constructed of a lightweight, high impact plastic case with a clear viewing window. The sealed front panel meets NEMA 4X/IP65 specifications for wash-down and/or dusty environments, when properly installed. A CUB4V or CUB4I unit can be mounted in the same panel cutout as the CUBVD and CUBID units.
The optional Micro Line/Sensor Power Supply (MLPS1000) is designed to attach to the rear of an installed CUB4V or CUB4I to provide the necessary power for the unit. The optional supply can be powered from either a 115 or 230 VAC source.
$173.00 - $189.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Display Digits Max DC Voltage VDC (VDC) Description
Availability Pricing
Red Lion CUB4I000 DC Current Panel Meter, Reflective EW-68484-28 Mfr # CUB4I000
COMPARE
3.5 199.9 DC Current Panel Meter, Reflective
$173.00USD / Each
Red Lion CUB4I010 DC Current Panel Meter, Yellow/Green, Backlit EW-68484-29 Mfr # CUB4I010
COMPARE
3.5 199.9 DC Current Panel Meter, Yellow/Green, Backlit
$189.00USD / Each
Red Lion CUB4I020 Dc Currmeter W/R Ed Backlighting EW-68484-30 Mfr # CUB4I020
COMPARE
3.5 199.9 Dc Currmeter W/R Ed Backlighting
$189.00USD / Each
Red Lion CUB4V000 DC Voltage Panel Meter, Reflective EW-68484-40 Mfr # CUB4V000
COMPARE
3.5 199.9 DC Voltage Panel Meter, Reflective
$173.00USD / Each
Red Lion CUB4V010 DC Voltage Panel Meter, Backlit, Yellow/Green EW-68484-41 Mfr # CUB4V010
COMPARE
3.5 199.9 DC Voltage Panel Meter, Backlit, Yellow/Green
$189.00USD / Each
Red Lion CUB4V020 DC Voltage Panel Meter, Backlit, Red EW-68484-42 Mfr # CUB4V020
COMPARE
3.5 199.9 DC Voltage Panel Meter, Backlit, Red
$189.00USD / Each
Showing 1- 6 of 6
Need Help with this product?
Call Us At1-847-549-7600
Live chat