Red Lion Miniature Process -Loop Powered Indicators
  • FEATURES
  • Dual range, 4 to 20mA or 10 to 50 mA
  • 3-1/2 digit, 0.6" high digits
  • Positive image transflective LCD with red backlight or positive image reflective LCD (CUB4LP)
  • Positive image transflective LCD with red backlight or negative image transmissive with red or yellow / green backlight (CUB4CL)
  • ,li> Span and offset capability
  • Negative and overrange indication
  • Selectable decimal point position
  • NEMA 4X / IP65 sealed front panel Bezel
  • Fits DIN standard cutout 2.68" x 1.30"
  • UL, CUL and CE approved
The CUB4LP uses a 4 to 20 mA or a 10 to 50 mA input signal as operating power. The input signal is also used to power the backlighting on the CUB4LP40 unit. The CUB4CL uses a 4 to 20 mA or a 10 to 50 mA input signal to power the unit.
An external power supply is used to power the CUB4CL backlighting to provide a brighter, more consistent display and a lower compliance voltage.
The units have a 3 1/2-digit LCD display with 0.6" (15.2 mm) high digits and a DIP switch selectable decimal point. The CUB4LP display is available in positive image reflective (dark digits, reflective background) or positive image transflective (dark digits, illuminated background) with red backlighting. The CUB4CL display is available in positive image transflective (dark digits, illuminated background) with red or yellow/green backlighting or negative image transmissive (illuminated digits, dark background) with red or yellow/green backlighting.
The ability to scale the display allows indication in any desired unit of measurement such as temperature, pressure, humidity, fluid flow, etc. The unit is calibrated at the factory with 0.0 displayed @ 4 mA input and 100.0 displayed @ 20 mA input.
The units are contained in a lightweight, high impact plastic case with a clear viewing window. When properly installed, the sealed front panel meets NEMA 4X/IP65 specifications for wash-down and dusty environments.
$112.00 - $254.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
DIN Size Input Type Max DC Voltage VDC (VDC)
Availability Pricing
Red Lion CUB4CL10 Current loop powered process indicator, with negative backlight EW-68484-24 Mfr # CUB4CL10
COMPARE
$254.00USD / Each
Red Lion CUB4CL20 Current Loop W/Red Neg. Backlight EW-68484-25 Mfr # CUB4CL20
COMPARE
$254.00USD / Each
Red Lion CUB4CL30 Current Loop W/Yel /Grn Pos. Backlight EW-68484-26 Mfr # CUB4CL30
COMPARE
$254.00USD / Each
Red Lion CUB4CL40 Current Loop W/Red Pos. Backlight EW-68484-27 Mfr # CUB4CL40
COMPARE
$254.00USD / Each
Red Lion CUB4L000 CUB 4 COUNTERREFLECTIVE EW-68484-31 Mfr # CUB4L000
COMPARE
$112.00USD / Each
Red Lion CUB4LP00 Miniature Loop-Powered Process Indicator, Reflective LCD EW-68484-37 Mfr # CUB4LP00
COMPARE
$238.00USD / Each
Red Lion CUB4LP40 Miniature Loop-Powered Process Indicator, Transflective LCD EW-68484-38 Mfr # CUB4LP40
COMPARE
$242.00USD / Each
Red Lion CUB5PB00 PROCESS METERW/ RED/GRN BKLT EW-68864-85 Mfr # CUB5PB00
COMPARE
$207.00USD / Each
Red Lion CUB5TR00 COUNTERCYCLEREFLECTIVE EW-68864-86 Mfr # CUB5TR00
COMPARE
$152.00USD / Each
Showing 1- 9 of 9
Need Help with this product?
Call Us At1-847-549-7600
Live chat