This product is now discontinued, View Related Products
Red Lion Process Control Units
FEATURES
  • Accepts different types of process inputs (0 to 10VDC, 0/4 to 20mA)
  • Full PID control with reduced overshoot
  • Status Indicators for outputs and control modes
  • Programmable user input (digital) for added flexibility
  • Manual/Automatic and local/remote setpoint control modes
  • Setpoint ramping for process start-up
  • Field replaceable and interchangeable output modules (Relay, Logic/SSR Drive and Triac)
  • 100 msec sampling period with 0.15% accuracy
  • On demand auto-tuning of PID control settings
  • Dual LED displays for simultaneous indication of process value and setpoint or second analog input
* Note: Output modules ARE NOT supplied with the controller. All output modules are packaged separately and must be installed by the user. When selecting the controller, be sure to purchase the appropriate output module for the main control output and if necessary, the alarm output(s), the cooling output, and valve positioner outputs. The controller can be fitted with any combination of output modules. The logic/SSR Drive Module is a switched DC source, intended to drive the DC input of an SSR power unit. It should never be connected to line voltage.
Call For Price
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
Red Lion PCU10000 1/8 Din Process Control Unit Base Unit EW-68486-29 Mfr # PCU10000
COMPARE
1/8
Call For Price
Red Lion PCU10001 Process Control Unit with Alarm, NEMA 4X EW-68486-30 Mfr # PCU10001
COMPARE
1/8
Call For Price
Red Lion PCU10002 W/ Nema 4X & Secon Dary Output EW-68486-31 Mfr # PCU10002
COMPARE
1/8
Call For Price
Red Lion PCU10104 Process Control Unit with Alarm, NEMA 4X, RS-485 EW-68486-32 Mfr # PCU10104
COMPARE
1/8
Call For Price
Red Lion PCU11001 Process Control Unit, NEMA 4X EW-68486-34 Mfr # PCU11001
COMPARE
1/8
Call For Price
Red Lion PCU11002 Process Control Unit, NEMA 4X, Analog and Secondary EW-68486-35 Mfr # PCU11002
COMPARE
1/8
Call For Price
Red Lion PCU11004 1/8 Din Process Control Unit Analog Alarm And Rs-485 Output EW-68486-36 Mfr # PCU11004
COMPARE
1/8
Call For Price
Red Lion PCU11005 W/ Nema 4X Anlg Secondary & Rs485 EW-68486-37 Mfr # PCU11005
COMPARE
1/8
Call For Price
Red Lion PCU11108 1/8 Din Process Control Unit Remote Setpoint 4/20 Analog And Alarm EW-68486-38 Mfr # PCU11108
COMPARE
1/8
Call For Price
Showing 1- 9 of 9