Red Lion Real Time Clock Elapsed Time Panel Meters
  • FEATURES
  • 1/8 DIN (50 mm x 97 mm)
  • Six digit LED display
  • Elapsed timer with preset capability
  • Four separate displays (timer, counter, real time clock and date)
  • PC software programmable
  • Four setpoint alarm outputs (with plug in card)
  • Communications and bus capabilities (with plug in card)
  • NEMA 4X / IP65 front bezel
The PAXTM (Timer) and PAXCK (Clock/Timer) can both function as an elapsed time or preset timer, which the PAXCK also offers additional real-time clock with date capability. By using two separate signal inputs and 23 selectable time ranges, the meter can be programmed to meet most any timing application. With the addition of a plug-in setpoint card, the PAXCK can be utilized as a dual or quad output preset timer.
The PAXCK can operate as a Real-Time Clock with option card PAXRTC00. The meter is capable of displaying time in 12-hour or 24-hour time formats. The meter is also capable of displaying day, month and year. The meter will recognize leap year and automatically adjust for Daylight Savings Time.
$244.00 - $345.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
DIN Size Input Type Max DC Voltage VDC (VDC)
Availability Pricing
Red Lion PAXCK000 85-250AC 1/8 Din Real Time Clock Panel Meter EW-68485-97 Mfr # PAXCK000
COMPARE
1/8"
$303.00USD / Each
Red Lion PAXCK010 11-36VDC/24Vac 1/8 Din Real Time Clock Panel Meter EW-68485-98 Mfr # PAXCK010
COMPARE
1/8"
$345.00USD / Each
Red Lion PAXTM000 1/8 Din Elapsed/Preset Timer 85-250Vac EW-68486-22 Mfr # PAXTM000
COMPARE
1/8"
$244.00USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat