This product is now discontinued, View Related Products
Red Lion Selectable - Timebase Tachometer
The DT8 is a self-powered rate indicator which features selectabletimebase increments by setting the appropriate DIP switches on the rear of the unit. The internal 3.0 VDC lithium battery will operate continuously for at least 7 years. It has a 5-digit LCD display with 0.6 inch (15.2mm) high digits.
The unit is constructed of a lightweight, high impact plastic case with a clear viewing window. The sealed front panel meets NEMA 4X/IP65 specifications for wash-down and/or dusty environments, when properly installed. The optional Micro Line/Sensor Power Supply (RLMLPS10000) is designed to attach to the rear of an installed DT8. The optional supply can be powered from an 85 to 250 VAC source, and can provide power for the backlighting of a unit and most sensors.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Display Type Max Power VDC (VDC) Description
Availability Pricing
Red Lion DT800000 Adjustable Time Base Tachometer EW-68484-60 Mfr # DT800000
COMPARE
0.6\" backlit LCD 28 Adjustable Time Base Tachometer
Call For Price
Red Lion DT800010 Adj Timebase Tach W/Red Backlighting EW-68484-61 Mfr # DT800010
COMPARE
0.6\" backlit LCD 28 Adj Timebase Tach W/Red Backlighting
Call For Price
Red Lion DT800020 Adj.Timebase Tachometer W/Red Backlighting EW-68484-62 Mfr # DT800020
COMPARE
0.6\" backlit LCD 28 Adj.Timebase Tachometer W/Red Backlighting