Reduced-Pulsation Diaphragm Metering Pumps
Chemical resistance prevents contamination of your pumped medium
  • Quiet, reliable, accurate, and virtually pulse-free
  • Pumps can run dry continuously without damage
  • Built-in overpressure protection prevents damage to pump
A convenient way to transfer liquids in the laboratory. Compact and resistant to chemical attack, these pumps prevent contamination of your pumped medium. Larger pump head geometry and wetted materials provide the longest possible lifetime when pumping corrosive liquids, gases, and vapors. Pumps can have either local or external contact capabilities—external protocols include 0 to 10 VDC, start/via TTL contact, and output fault alarm.
$1,173.00 - $4,162.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Inlet Connection Max Flow Rate (mL/min) Power (VAC)
Availability Pricing
KNF FEM 08KT.18S Manual Control Reduced Pulsation Diaphragm Metering Pump, 90 psi EW-78166-40 Mfr # NF1.100KT.18S
COMPARE
5/16" hose barb 1300 100 to 230
$1,173.00USD / Each
KNF FEM 08KT.18S Diaphragm Metering Pump, PP head, 500 to 3000 mL/min flow range, 90 psi EW-78166-41 Mfr # NF1.300KT.18S
COMPARE
1/2" hose barb 1300 100 to 230
$1,377.00USD / Each
KNF FEM 08KT.18S Diaphragm Metering Pump, PVDF head, 200 to 1300 mL/min flow range, 90 psi EW-78166-42 Mfr # NF1.100TT.18S
COMPARE
5/16" hose barb 1300 100 to 230
$1,397.00USD / Each
KNF FEM 08KT.18S Diaphragm Metering Pump, PVDF head, 500 to 3000 mL/min flow range, 90 psi EW-78166-43 Mfr # NF1.300TT.18S
COMPARE
1/2" hose barb 3000 100 to 230
$1,266.00USD / Each
KNF FEM 08KT.18S Diaphragm Metering Pump, PTFE head, 200 to 1300 mL/min flow range, 90 psi EW-78166-44 Mfr # NF1.100FT.18S
COMPARE
5/16" hose barb 1300 100 to 230
$3,163.00USD / Each
KNF FEM 08KT.18S Diaphragm Metering Pump, PTFE head, 500 to 3000 mL/min flow range, 90 psi EW-78166-45 Mfr # NF1.300FT.18S
COMPARE
1/2" hose barb 3000 100 to 230
$4,162.00USD / Each