This product is now discontinued, View Related Products
Refrigeration Dryers
Eliminate damage-causing moisture from your compressed air lines
  • “Plug and play” design for easy installation and operation
  • Low pressure differential across the dryer (1.45 psig on average)
  • Environmentally friendly refrigerant
  • Oversized, high-efficiency condenser to operate in ambients to 122°F (50°C)
  • Maintenance-free, hermetically sealed refrigerant compressor
  • All models incorporate a dew point indicator and level-sensing automatic float drain
Parkers'' Starlette Plus refrigeration dryers eliminate damage causing moisture from your compressed air lines. Active removal of condensate lets you achieve near-perfect dry compressed air—resulting in less downtime, reduced costs and maintenance, and an improved finished product. This new dryer design offers the most compact dimensions in the marketplace, making it easy to integrate in your existing systems.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Free Air Capacity (CFM) Max Pressure (PSI) Max Vacuum
Availability Pricing
Parker Hannifin Refrigeration Dryer for Compressors, 5 SCFM nominal flow, 115 VAC, 60 Hz EW-79162-60 Mfr # BRD5
COMPARE
Call For Price
Parker Hannifin Refrigeration Dryer for Compressors, 10 SCFM nominal flow, 115 VAC, 60 Hz EW-79162-61 Mfr # SPE010-A11516016TIU
COMPARE
Call For Price
Parker Hannifin BRD15 Refrigeration Dryer for Compressors, 15 SCFM nominal flow, 115 VAC, 60 Hz EW-79162-62 Mfr # BRD15
COMPARE
Call For Price
Parker Hannifin BRD25 Refrigeration Dryer for Compressors, 25 SCFM nominal flow, 115 VAC, 60 Hz EW-79162-63 Mfr # BRD25
COMPARE
Call For Price
Parker Hannifin BRD35 Refrigeration Dryer for Compressors, 35 SCFM nominal flow, 115 VAC, 60 Hz EW-79162-64 Mfr # BRD35
COMPARE
Call For Price
Parker Hannifin BRD50 Refrigeration Dryer for Compressors, 50 SCFM nominal flow, 115 VAC, 60 Hz EW-79162-65 Mfr # BRD50
COMPARE
Call For Price
Parker Hannifin BRD75 Refrigeration Dryer for Compressors, 75 SCFM nominal flow, 115 VAC, 60 Hz EW-79162-66 Mfr # BRD75
COMPARE
Call For Price
Showing 1- 7 of 7