Remote-Control Diaphragm Liquid Dosing Pumps
Uniformity of discharge ensures low-pulsation transfer process
  • Quiet, reliable, accurate, and virtually maintenance-free
  • Pumps can run dry continuously without damage
  • Versatile pumps handle flow rates up to 100 mL/min and volumes up to 1000 mL
Simple and intuitive function makes the liquid dosing pump an accurate and reliable solution for your laboratory dosing applications. User-friendly design provides a quiet, dry-running, and self-priming dosing operation. Easy calibration and cycle mode allow for uninterrupted and virtually maintenance-free dosing.
This pump features a PTFE-covered diaphragm and IP65-rated splash protection, making it resistant to chemical attack and adaptable to fluid characteristics.
Unique drive system ensures maximum uniformity of discharge resulting in a low-pulsation transfer process. Pumps feature either local or external control capabilities including start/stop and reset/prime via TTL contact as well as output signal fault alarm. Models available with a RS-232 port for connectivity.
$2,123.00 - $5,171.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Wetted Materials
Availability Pricing
KNF FEM 08KT.18S Digital Control Dosing Metering Pump, PP head, 0.03 to 20 mL/min flow range EW-78167-00 Mfr # FEM1.02KT.18RC
COMPARE
20 85 Polypropylene head, PTFE diaphragm, FFKM valves
$2,153.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Dosing Pump, PVDF head, 0.03 to 20 mL/min EW-78167-01 Mfr # FEM1.02TT.18RC
COMPARE
20 85 PVDF head, PTFE diaphragm, FFKM valves
$2,285.00USD / Each
KNF FEM 08KT.18S Digital Control Dosing Metering Pump, PTFE head, 0.03 to 20 mL/min EW-78167-02 Mfr # FEM1.02FT.18RC
COMPARE
20 85 PTFE head, PTFE diaphragm, FFKM valves
$3,754.00USD / Each
KNF FEM 08KT.18S Digital Control Dosing Metering Pump, SS head, 0.03 to 20 mL/min EW-78167-03 Mfr # FEM1.02ST.18RC
COMPARE
20 85 Stainless Steel head, PTFE diaphragm, FFKM valves
$4,804.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Plus Dosing Pump, PP head, 0.03 to 20 mL/min EW-78167-04 Mfr # FEM1.02KT.18RCP
COMPARE
20 85 Polypropylene head, PTFE diaphragm, FFKM valves
$2,520.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Plus Dosing Pump, PVDF head, 0.03 to 20 mL/min EW-78167-05 Mfr # FEM1.02TT.18RCP
COMPARE
20 85 PVDF head, PTFE diaphragm, FFKM valves
$2,754.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Plus Dosing Pump, PTFE head, 0.03 to 20 mL/min EW-78167-06 Mfr # FEM1.02FT.18RCP
COMPARE
20 85 PTFE head, PTFE diaphragm, FFKM valves
$4,121.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Plus Dosing Pump, SS head, 0.03 to 20 mL/min EW-78167-07 Mfr # FEM1.02ST.18RCP
COMPARE
20 85 Stainless Steel head, PTFE diaphragm, FFKM valves
$5,171.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Dosing Pump, PP head, 1 to 100 mL/min EW-78167-08 Mfr # FEM1.10KT.18RC
COMPARE
100 85 Polypropylene head, PTFE diaphragm, FFKM valves
$2,123.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Dosing Pump, PVDF head, 1 to 100 mL/min EW-78167-09 Mfr # FEM1.10TT.18RC
COMPARE
100 85 PVDF head, PTFE diaphragm, FFKM valves
$2,235.00USD / Each
KNF FEM 08KT.18S Digital Remote Control Dosing Pump, PTFE head, 1 to 100 mL/min EW-78167-10 Mfr # FEM1.10FT.18RC
COMPARE
100 85 PTFE head, PTFE diaphragm, FFKM valves
$3,683.00USD / Each
Showing 1- 11 of 11