Replacement PFA Compression Ferrule Nut and Assemblies
Create a secure connection with compression fittings
  • Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
$20.00 - $97.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Parker Hannifin Compression Ferrule Plug Fitting, PFA, 1/8"; 1/Each
Buy More Save More
EW-31320-45 Mfr # GTPG-2
COMPARE
1/8 PFA Ferrule plug, PFA, 1/8"
$82.00USD / Each
Parker Hannifin Compression Ferrule Plug Fitting, PFA, 1/4"; 1/Each
Buy More Save More
EW-31320-46 Mfr # GTPG-4
COMPARE
1/4 PFA Ferrule plug, PFA, 1/4"
$65.50USD / Each
Parker Hannifin Compression Ferrule Plug Fitting, PFA, 3/8"; 1/Each
Buy More Save More
EW-31320-47 Mfr # GTPG-6
COMPARE
3/8 PFA Ferrule plug, PFA, 3/8"
$87.00USD / Each
Parker Hannifin Compression Ferrule Plug Fitting, PFA, 1/2"; 1/Each
Buy More Save More
EW-31320-48 Mfr # GTPG-8
COMPARE
1/2 PFA Ferrule plug, PFA, 1/2"
$90.00USD / Each
Parker Hannifin Compression Ferrule Plug Fitting, PFA, 3/4"; 1/Each
Buy More Save More
EW-31320-49 Mfr # GTPG-12
COMPARE
3/4 PFA Ferrule plug, PFA, 3/4"
$97.00USD / Each
Parker Hannifin Fitting, PFA, Compression Nut Assembly, 1/8" OD
Buy More Save More
EW-31321-85 Mfr # 1202-0012
COMPARE
1/8 PFA Compression Nut Assembly, PFA, 1/8"
$20.00USD / Each
Parker Hannifin Fitting, PFA, Compression Nut Assembly, 1/4" OD
Buy More Save More
EW-31321-86 Mfr # 1202-0001
COMPARE
1/4 PFA Compression Nut Assembly, PFA, 1/4"
$20.00USD / Each
Parker Hannifin Fitting, PFA, Compression Nut Assembly, 3/8" OD
Buy More Save More
EW-31321-87 Mfr # 1202-0002
COMPARE
3/8 PFA Compression Nut Assembly, PFA, 3/8"
$24.00USD / Each
Parker Hannifin Fitting, PFA, Compression Nut Assembly, 1/2" OD
Buy More Save More
EW-31321-88 Mfr # 1202-0003
COMPARE
1/2 PFA Compression Nut Assembly, PFA, 1/2"
$26.00USD / Each
Parker Hannifin Fitting, PFA, Compression Nut Assembly, 3/4" OD
Buy More Save More
EW-31321-89 Mfr # 1202-0004
COMPARE
3/4 PFA Compression Nut Assembly, PFA, 3/4"
$34.50USD / Each
Showing 1- 10 of 10