ResinTech Phoenix UHP Water System Accessories and Options
Accessories, options, and consumables for Phoenix Academy, Citation, Biochem, and Genome Ultra-hihg Purity Water Systems
A complete package requires a system, conditioning cartridge, polishing cartridge, and final filter.
1. Choose from four base Phoenix systems with benchtop or remote dispensing
2. Choose conditioning cartridge 99285-45 for RO water feed or 99285-46 for SDI/tap water feed
3. Choose polishing cartridge 99285-43 for standard results or 99285-44 for additional organics removal
4. Choose final filter 99285-27 for 0.2µm pariculate removal or 99285-47 for additional endotoxin removal
Options (must be ordered with unit, factory installation required):
Item 99285-39 - adds in flow totalizer function, viewable on display
Item 99285-40 - adds in a direct feed port
Item 99285-41 - adds in a recirculating dispensing gun (for units without a remote dispense function)
Item 99285-42 - adds in a wall mounting bracket to clear up bench space
Consumables (depend upon the base system chosen):
Item 99285-48 is the replacement ultrafilter for the Biochem and Genome models
Item 99285-49 is the replacement UV lamp for the Citation and Genome models
And item 99285-50 is the sanitatino skit to be used by all Phoenix models (Academy, Biochem, Citation, and Genome)
$167.25 - $1,227.50USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description Special Features
Availability Pricing
ResinTech Phoenix Direct Feed Port EW-99285-40 Mfr # CLA-5000-DF
COMPARE
Direct Feed Port Optional
$237.00USD / Each
ResinTech Phoenix Recirculating Dispensing Gun with 0.2 micron filter EW-99285-41 Mfr # CLA-5000-DG
COMPARE
Recirculating Dispensing Gun with 0.2 micron filter Optional
$1,227.50USD / Each
ResinTech Phoenix Wall Mounting Bracket EW-99285-42 Mfr # CLA-5000-WB
COMPARE
Wall Mounting Bracket Optional
$167.25USD / Each
ResinTech Phoenix Polishing Ultrapure Cartridge EW-99285-43 Mfr # CLF-5000-P1
COMPARE
Polishing Ultrapure Cartridge Requirement/Consumable
$238.75USD / Each
ResinTech Phoenix Polishing Ultrapure Low Organics Cartridge EW-99285-44 Mfr # CLF-5000-P2
COMPARE
Polishing Ultrapure Low Organics Cartridge Requirement
$277.50USD / Each
ResinTech Phoenix Dual Conditioning Pack, RO Feed EW-99285-45 Mfr # CLF-5000-C1
COMPARE
Dual Conditioning Pack, RO Feed Requirement
$450.50USD / Each
ResinTech Phoenix Dual Conditioning Pack, SDI/TAP Feed EW-99285-46 Mfr # CLF-5000-C2
COMPARE
Dual Conditioning Pack, SDI/TAP Feed Requirement
$463.25USD / Each
ResinTech Phoenix 0.2 Micron Capsule Endotoxin Filter EW-99285-47 Mfr # CLF-5000-02E
COMPARE
0.2 Micron Capsule Endotoxin Filter Requirement
$444.25USD / Each
ResinTech Phoenix Ultrafilter Replacement, 3/8" Quick Disconnect Fitting EW-99285-48 Mfr # PF-00-7105
COMPARE
Ultrafilter Replacement, 3/8" Quick Disconnect Fitting Consumable
$594.75USD / Each
ResinTech Phoenix UV Bulb Replacement EW-99285-49 Mfr # CLX-5000-016
COMPARE
UV Bulb Replacement Consumable
$239.00USD / Each
ResinTech Phoenix Sanitization Kit EW-99285-50 Mfr # CLX-5000-017
COMPARE
Sanitization Kit Consumable
$222.00USD / Each
Showing 1- 11 of 11