This product is now discontinued, View Related Products

Retsch Cryogenic Jar Mill Accessories

Accessories for your Retsch cryomill
The Retsch connection tube and autofill container are designed for use with the Retsch CryoMill. The connection tube comes included with safety valve. The autofill system avoids direct contact with LN2 and makes cryogenic grinding very safe.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Retsch Autofill Container Ln2 10l For Cryomill EW-04182-25 Mfr # 02.480.0001
COMPARE
Autofill Container Ln2 10l For Cryomill
Call For Price
Retsch 05.871.0001 Connection Tube For Ln2 Tank EW-04182-26 Mfr # 05.871.0001
COMPARE
Connection Tube For Ln2 Tank
Call For Price
Retsch 02.706.0272 Cryogenic Jar Mill Jar Adapter; 5 mL EW-04182-27 Mfr # 02.706.0272
COMPARE
Cryogenic Jar Mill Jar Adapter; 5 mL
Call For Price
Retsch 02.480.0002 Autofill Container Ln2 50l For Cryomill EW-04182-28 Mfr # 02.480.0002
COMPARE
Autofill Container Ln2 50l For Cryomill
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat