RH Series Piston Pump Heads
Valveless piston pumps are low maintenance and suffer fewer piston siezures or clogs. With a single moving part, there is less drift over time, offering millions of cycles of reliable, repeatable pumping.
$611.25 - $749.50USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Inlet Connection Max Flow Rate (mL/min) Wetted Materials
Availability Pricing
FMI Series RH Valveless Piston Pump Head, Stainless Steel/PVDF/Carbon; 0 to 0.025 mL per Stroke EW-07117-33 Mfr # FMI002 / RH00SKY
COMPARE
1/4" OD tubing 18.75 316 SS, PVDF, Carbon (sintered)
$675.75USD / Each
FMI Series RH Valveless Piston Pump Head, Stainless Steel/ETFE/Carbon; 0 to 0.025 mL per Stroke EW-07117-35 Mfr # FMI003 / RH00STY
COMPARE
1/4" OD tubing 18.75 316 SS, ETFE, Carbon (sintered)
$749.50USD / Each
FMI Series RH Valveless Piston Pump Head, Stainless Steel/ETFE/Carbon; 0 to 0.025 mL per Stroke, with 1/4-28 NPT(F) Ports EW-07117-41 Mfr # FMI011 / RH00STY-LF
COMPARE
1/4-28 female 18.75 316 SS, ETFE, Carbon (sintered)
$727.25USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic/ETFE/Ceramic; 0 to 0.05 mL per Stroke EW-07117-53 Mfr # FMI006 / RH0CTC
COMPARE
1/4" OD tubing 37.5 Ceramic (alumina), ETFE
$702.50USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic/PVDF/Ceramic; 0 to 0.05 mL per Stroke EW-07117-55 Mfr # FMI005A / RH0CKC
COMPARE
1/4" OD tubing 37.5 Ceramic (alumina), PVDF
$611.25USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic/PVDF/Ceramic; 0 to 0.05 mL per Stroke, with 1/4-28 NPT(F) Ports EW-07117-59 Mfr # FMI013 / RH0CKC-LF
COMPARE
1/4-28 female 37.5 Ceramic (alumina), PVDF
$625.50USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic/ETFE/Ceramic; 0 to 0.05 mL per Stroke, with 1/4-28 NPT(F) Ports EW-07117-61 Mfr # FMI014 / RH0CTC-LF
COMPARE
1/4-28 female 37.5 Ceramic (alumina), ETFE
$687.75USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic/ETFE/Ceramic; 0 to 0.10 mL per Stroke EW-07117-63 Mfr # FMI008A / RH1CTC
COMPARE
1/4" OD tubing 75 Ceramic (alumina), ETFE
$686.00USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic/PVDF/Ceramic; 0 to 0.10 mL per Stroke EW-07117-65 Mfr # RH1CKC
COMPARE
1/4" OD tubing 75 Ceramic (alumina), PVDF
$619.50USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic, 1/4", 1/4-28 (F) ports EW-07117-69 Mfr # RH1CKCLF
COMPARE
1/4-28 female 75 Ceramic (alumina), PVDF
$613.50USD / Each
FMI Series RH Valveless Piston Pump Head, Ceramic/ETFE/Ceramic; 0 to 0.10 mL per Stroke, with 1/4-28 UNF (F) ports EW-07117-71 Mfr # FMI016 / RH1CTC-LF
COMPARE
1/4-28 female 75 Ceramic (alumina), ETFE
$673.00USD / Each
Showing 1- 11 of 11