Ribonuclease A (From Bovine Pancreas)

  • Description - White Lyophilized Powder
  • Activity - ≥50 Kuntz units/mg
$202.00USD / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical BP2539100 Ribonuclease A (From Bovine Pancreas) (100mg) EW-88060-70 Mfr # BP2539100
COMPARE
9001-99-4
$202.00USD / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat