Ricca Chemical Company Ammonia Standard
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$42.60 - $277.25USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
Ricca Chemical Company 612-16 Ammonia Standard, 1 mL = 0.01 mg NH3, 10 ppm NH3 (8.22 ppm N), 500 mL EW-86969-45 Mfr # 612-16
COMPARE
500 Ammonia Standard, 1 mL = 0.01 mg NH3, 10 ppm NH3 (8.22 ppm N), 500 mL
$83.40USD / Each
Ricca Chemical Company 613-16 Ammonia Standard, 1 mL = 0.1 mg NH3, 100 ppm NH3 (82.2 ppm N), 500 mL EW-86969-46 Mfr # 613-16
COMPARE
500 Ammonia Standard, 1 mL = 0.1 mg NH3, 100 ppm NH3 (82.2 ppm N), 500 mL
$78.80USD / Each
Ricca Chemical Company 615-16 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 500 mL EW-86969-47 Mfr # 615-16
COMPARE
500 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 500 mL
$84.00USD / Each
Ricca Chemical Company 613-32 Ammonia Standard, 1 mL = 0.1 mg NH3, 100 ppm NH3 (82.2 ppm N), 1L EW-86977-84 Mfr # 613-32
COMPARE
1000 Ammonia Standard, 1 mL = 0.1 mg NH3, 100 ppm NH3 (82.2 ppm N), 1L
$131.25USD / Each
Ricca Chemical Company 615-32 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 1L EW-86977-85 Mfr # 615-32
COMPARE
1000 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 1L
$143.75USD / Each
Ricca Chemical Company 615-1 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 4L EW-86984-75 Mfr # 615-1
COMPARE
3785 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 4L
$277.25USD / Each
Ricca Chemical Company 615-4 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 120 mL EW-86989-38 Mfr # 615-4
COMPARE
120 Ammonia Standard, 1 mL = 1 mg NH, 1000 ppm NH3 (822 ppm N), 120 mL
$42.60USD / Each
Showing 1- 7 of 7