Ricca Chemical Company® HydroSpec™ Volumetric Reagents for Karl Fischer Titrations

Specially formulated reagents ensure reliable and reproducible results
  • Support a wide range of sample types from ethanol and fuels to fats and oils
  • Available for both one- and two-component titrations
These highly stable solvents retain activity for up to 5 years. 1 L bottles have a GL45 threaded neck to make connection to Karl Fischer titrators easy and simple.
$112.25 - $122.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Ricca Chemical Company HydroSpec™ Titrant T5 General Use Two Component Volumetric Solution; 1 L EW-86994-90 Mfr # RK230000-1C
COMPARE
Includes 67-56-1, 7553-56-2 1000 Titrant T5 General Use Two Component Volumetric Solution; 1 L
$122.00USD / Each
Ricca Chemical Company HydroSpec™ Titrant T5 General Use Two Component Volumetric Solution; 6 x 1 L EW-86994-92 Mfr # RK230000-6x1C
COMPARE
Includes 67-56-1, 7553-56-2 6000 Titrant T5 General Use Two Component Volumetric Solution; 6 x 1 L
Call For Price
Ricca Chemical Company HydroSpec™ Titrant T2  Low Moisture Two Component Volumetric Solution; 1 L EW-86998-31 Mfr # RK250000-1C
COMPARE
Includes 67-56-1, 7553-56-2 1000 Titrant T2 Low Moisture Two Component Volumetric Solution; 1 L
$112.25USD / Each
Ricca Chemical Company HydroSpec™ Titrant T2  Low Moisture Two Component Volumetric Solution; 6 x 1 L EW-86998-33 Mfr # RK250000-6x1C
COMPARE
Includes 67-56-1, 7553-56-2 6000 Titrant T2 Low Moisture Two Component Volumetric Solution; 6 x 1 L
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat