Ricca Chemical Company Nitrate Standard

High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$40.10 - $84.80USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Form Volume (mL)
Availability Pricing
Ricca Chemical Company 5307-16 Nitrate Standard, 1 mL = 1 mg NO3-, 1000 ppm (500 mL) EW-86973-86 Mfr # 5307-16
COMPARE
Includes 7757-79-1 and 7732-18-5 Liquid 500
$84.80USD / Each
Ricca Chemical Company 5307-4 Nitrate Standard, 1 mL = 1 mg NO3-, 1000 ppm (120 mL) EW-86991-47 Mfr # 5307-4
COMPARE
Includes 7757-79-1 and 7732-18-5 Liquid 120
$40.10USD / Each
Ricca Chemical Company 5445-4 Nitrite Standard, 1 mL = 1 mg, 1000 ppm (120 mL) EW-86991-49 Mfr # 5445-4
COMPARE
Includes 67-66-3, 7632-00-0 and 7732-18-5 Liquid 120
$79.70USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat