Ricca Chemical Company Silver Nitrate Solutions
High quality, competitive pricing, and rapid service
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$41.30 - $997.00USD / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
Ricca Chemical Company 6825.7-16 Silver Nitrate, 1.7% (w/v) Aqueous Solution (17 g/L) (500 mL) EW-86975-49 Mfr # 6825.7-16
COMPARE
500 Silver Nitrate, 1.7% (w/v) Aqueous Solution (17 g/L) (500 mL)
$65.70USD / Each
Ricca Chemical Company Silver Nitrate, 5% (w/v) Aqueous Solution; 500 mL EW-86975-50 Mfr # 6828-16
COMPARE
500 Silver Nitrate, 5% (w/v) Aqueous Solution (500 mL)
$151.50USD / Each
Ricca Chemical Company 6850-16 Silver Nitrate, 0.0100 Normal (N/100) (500 mL) EW-86975-52 Mfr # 6850-16
COMPARE
500 Silver Nitrate, 0.0100 Normal (N/100) (500 mL)
$45.10USD / Each
Ricca Chemical Company 7025-16 Silver Nitrate, 1.00 Normal (500 mL) (Liquid) EW-86975-56 Mfr # 7025-16
COMPARE
500 Silver Nitrate, 1.00 Normal (500 mL)
$742.00USD / Each
Ricca Chemical Company 6850-32 Silver Nitrate, 0.0100 Normal (N/100) (1 L) EW-86982-24 Mfr # 6850-32
COMPARE
1000 Silver Nitrate, 0.0100 Normal (N/100) (1 L)
$68.20USD / Each
Ricca Chemical Company 6860-32 Silver Nitrate, 0.0141 Normal (0.0141 Molar), 1 mL = 0.5 mg Cl- (1 L) EW-86982-25 Mfr # 6860-32
COMPARE
1000 Silver Nitrate, 0.0141 Normal (0.0141 Molar), 1 mL = 0.5 mg Cl- (1 L)
$73.00USD / Each
Ricca Chemical Company 6920-32 Silver Nitrate, 0.0250 Normal (N/40) (1 L) EW-86982-29 Mfr # 6920-32
COMPARE
1000 Silver Nitrate, 0.0250 Normal (N/40) (1 L)
$88.30USD / Each
Ricca Chemical Company 6950-32 Silver Nitrate, 0.0282 Normal, 1 mL = 1 mg Cl- (1 L) EW-86982-30 Mfr # 6950-32
COMPARE
1000 Silver Nitrate, 0.0282 Normal, 1 mL = 1 mg Cl- (1 L)
$93.10USD / Each
Ricca Chemical Company 6970-32 Silver Nitrate, 0.0500 Normal (N/20) (1 L) EW-86982-31 Mfr # 6970-32
COMPARE
1000 Silver Nitrate, 0.0500 Normal (N/20) (1 L)
$123.00USD / Each
Ricca Chemical Company 7002-32 Silver Nitrate, 0.141 Normal, 1 mL = 5 mg Cl- (1 L) EW-86982-34 Mfr # 7002-32
COMPARE
1000 Silver Nitrate, 0.141 Normal, 1 mL = 5 mg Cl- (1 L)
$256.00USD / Each
Ricca Chemical Company 7003-32 Silver Nitrate, 0.171 Normal, 1 mL = 10 mg NaCl (1 L) EW-86982-35 Mfr # 7003-32
COMPARE
1000 Silver Nitrate, 0.171 Normal, 1 mL = 10 mg NaCl (1 L)
$314.00USD / Each
Ricca Chemical Company 7011-32 Silver Nitrate, 0.282 Normal, 1 mL = 10 mg Cl- (1 L) EW-86982-38 Mfr # 7011-32
COMPARE
1000 Silver Nitrate, 0.282 Normal, 1 mL = 10 mg Cl- (1 L)
$448.00USD / Each
Ricca Chemical Company 7026-32 Silver Nitrate, 0.0100 Normal (N/100) in Isopropyl Alcohol (1 L) EW-86982-41 Mfr # 7026-32
COMPARE
1000 Silver Nitrate, 0.0100 Normal (N/100) in Isopropyl Alcohol (1 L)
$202.75USD / Each
Ricca Chemical Company 6860-1 Silver Nitrate, 0.0141 Normal (0.0141 Molar), 1 mL = 0.5 mg Cl- (4 L) EW-86987-28 Mfr # 6860-1
COMPARE
4000 Silver Nitrate, 0.0141 Normal (0.0141 Molar), 1 mL = 0.5 mg Cl- (4 L)
$136.25USD / Each
Ricca Chemical Company 6970-1 Silver Nitrate, 0.0500 Normal (N/20) (4 L) EW-86987-34 Mfr # 6970-1
COMPARE
4000 Silver Nitrate, 0.0500 Normal (N/20) (4 L)
$185.75USD / Each
Ricca Chemical Company Silver Nitrate, 0.141 Normal, 1 mL = 5 mg Cl-; 4 L EW-86987-37 Mfr # 7002-1
COMPARE
4000 Silver Nitrate, 0.141 Normal, 1 mL = 5 mg Cl- (4 L)
$796.50USD / Each
Ricca Chemical Company 7003-1 Silver Nitrate, 0.171 Normal, 1 mL = 10 mg NaCl (4 L) EW-86987-38 Mfr # 7003-1
COMPARE
4000 Silver Nitrate, 0.171 Normal, 1 mL = 10 mg NaCl (4 L)
$975.00USD / Each
Ricca Chemical Company 7000-2.5 Silver Nitrate, 0.100 Normal (N/10) (10 L) EW-86989-05 Mfr # R7000000-10F
COMPARE
10000 Silver Nitrate, 0.100 Normal (N/10) (10 L)
$997.00USD / Each
Ricca Chemical Company 7000-4 Silver Nitrate, 0.100 Normal (N/10) (120 mL) EW-86992-11 Mfr # 7000-4
COMPARE
120 Silver Nitrate, 0.100 Normal (N/10) (120 mL)
$41.30USD / Each
Showing 1- 19 of 19