Ricca Chemical Company VeriSpec™ 19-Element Calibration Standard
Multi-element/ion calibration solutions
  • Made to Guide 34 Standards!
$279.00 - $379.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ ICP Calibration Standard 19; 100 mL EW-85012-16 Mfr # RV010709-100N
COMPARE
Ag 50 ppm, Al 100 ppm, B 15 ppm, Ba 5 ppm, Be 1 ppm, Bi 200 ppm, Cd 20 ppm, Co 20 ppm, Cr 25 ppm, Cu 20 ppm, Fe 15 ppm, Ga 150 ppm, In 200 ppm, Mn 5 ppm, Ni 50 ppm, Pb 200 ppm, Sr 1 ppm, Tl 400 ppm, and Zn 20 ppm 5% Nitric Acid 100
$279.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 19; 100 mL EW-85013-72 Mfr # RV010866-100N
COMPARE
Ag 50 ppm, Al 100 ppm, B 15 ppm, Ba 5 ppm, Be 1 ppm, Bi 200 ppm, Cd 20 ppm, Co 20 ppm, Cr 25 ppm, Cu 20 ppm, Fe 15 ppm, Ga 150 ppm, In 200 ppm, Mn 5 ppm, Ni 50 ppm, Pb 200 ppm, Sr 1 ppm, Tl 400 ppm, and Zn 20 ppm 5% Nitric Acid 100
$379.00USD / Each