This product is now discontinued, View Related Products
Ricca Chemical Company VeriSpec™ 22-Element Calibration Standard
Multi-element/ion calibration solutions
  • Made to Guide 34 Standards!
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ QC ICV Standard 7; 100 mL EW-85011-73 Mfr # RV010666-100N
COMPARE
Ag 10 ppm, Al 200 ppm, As 10 ppm, Ba 200 ppm, Be 5 ppm, Ca 5000 ppm, Cd 5 ppm, Co 50 ppm, Cr 10 ppm, Cu 25 ppm, Fe 100 ppm, K 5000 ppm, Mg 5000 ppm, Mn 15 ppm, Na 5000 ppm, Ni 40 ppm, Pb 5 ppm, Sb 60 ppm, Se 5 ppm, Tl 10 ppm, V 50 ppm, and Zn 20 ppm 5% Nitric Acid 100
Call For Price
Ricca Chemical Company VeriSpec™ CDRL Standard 22; 100 mL EW-85013-44 Mfr # RV010837-100N
COMPARE
Ag 1 ppm, Al 20 ppm, As 1 ppm, Ba 20 ppm, Be 0.5 ppm, Ca 500 ppm, Cd 0.5 ppm, Co 5 ppm, Cr 1 ppm, Cu 2.5 ppm, Fe 10 ppm, K 500 ppm, Mg 500 ppm, Mn 1.5 ppm, Na 500 ppm, Ni 4 ppm, Pb 0.3 ppm, Sb 6 ppm, Se 0.5 ppm, Tl 1 ppm, V 5 ppm, and Zn 2 ppm 5% Nitric Acid 100