Ricca Chemical Company VeriSpec™ 30-Element Calibration Standard
Multi-element/ion calibration solutions
  • Made to Guide 34 Standards!
$620.00 - $1,026.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ Multi-Element Standard 30, 1000 ppm Ca, 100 ppm AS/B/Be/Fe/Se/Zn, 10 ppm of each other; 100 mL EW-85012-20 Mfr # RV010713-100N
COMPARE
Ag 10 ppm, Al 10 ppm, As 100 ppm, B 100 ppm, Ba 10 ppm, Be 100 ppm, Bi 10 ppm, Ca 1000 ppm, Cd 10 ppm, Co 10 ppm, Cr 10 ppm, Cu 10 ppm, Fe 100 ppm, Ga 10 ppm, K 10 ppm, Li 10 ppm, Mg 10 ppm, Mn 10 ppm, Mo 10 ppm, Na 10 ppm, Ni 10 ppm, Pb 10 ppm, Rb 10 ppm, Se 100 ppm, Sr 10 ppm, Te 10 ppm, Tl 10 ppm, U 10 ppm, V 10 ppm, and Zn 100 ppm 5% Nitric Acid 100
$710.00USD / Each
Ricca Chemical Company VeriSpec™ Trace Metals in Water; 100 mL EW-85012-68 Mfr # RV010761-100N
COMPARE
Ag 0.001 ppm, Al 0.142 ppm, As 0.06 ppm, B 0.158 ppm, Ba 0.544 ppm, Be 0.014 ppm, Bi 0.014 ppm, Ca 3.2 ppm, Cd 0.007 ppm, Co 0.027 ppm, Cr 0.02 ppm, Cu 0.023 ppm, Fe 0.098 ppm, K 2 ppm, Li 0.017 ppm, Mg 8 ppm, Mn 0.039 ppm, Mo 0.121 ppm, Na 2.1 ppm, Ni 0.062 ppm, Pb 0.02 ppm, Rb 0.014 ppm, Re 0.113 ppm, Sb 0.058 ppm, Se 0.012 ppm, Sr 0.323 ppm, Te 0.001 ppm, Tl 0.007 ppm, V 0.038 ppm, and Zn 0.079 ppm 5% Nitric Acid 100
$1,026.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 30, 10 ppm of each; 100 mL EW-85013-71 Mfr # RV010865-100N
COMPARE
Ag 10 ppm, Al 10 ppm, As 10 ppm, Ba 10 ppm, Be 10 ppm, Bi 10 ppm, Ca 10 ppm, Cd 10 ppm, Co 10 ppm, Cr 10 ppm, Cs 10 ppm, Cu 10 ppm, Fe 10 ppm, Ga 10 ppm, Hg 10 ppm, In 10 ppm, K 10 ppm, Li 10 ppm, Mg 10 ppm, Mn 10 ppm, Na 10 ppm, Ni 10 ppm, Pb 10 ppm, Rb 10 ppm, Se 10 ppm, Sr 10 ppm, Tl 10 ppm, U 10 ppm, V 10 ppm, and Zn 10 ppm 5% Nitric Acid 100
$620.00USD / Each