Ricca Chemical Company VeriSpec™ 4-Element Calibration Standard
Multi-element/ion calibration solutions
  • Made to Guide 34 Standards!
$172.00 - $598.00USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ Multi-Element Standard 4, 5000 ppm Al/Ca/Mg, 2000 ppm Fe; 100 mL EW-85011-76 Mfr # RV010669-100N
COMPARE
Al 5000 ppm, Ca 5000 ppm, Fe 2000 ppm, and Mg 5000 ppm 5% Nitric Acid 100
$272.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 4, 100 ppm Mo/Sb/Sn/Ti; 100 mL EW-85011-82 Mfr # RV010675-100N
COMPARE
Mo 100 ppm, Sb 100 ppm, Sn 100 ppm, and Ti 100 ppm 2% Nitric Acid and 0.5% Hydrofluoric Acid 100
$267.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 4, 5000 ppm Ca/K/Mg/Na; 100 mL EW-85012-13 Mfr # RV010706-100N
COMPARE
Ca 5000 ppm, K 5000 ppm, Mg 5000 ppm, and Na 5000 ppm 5% Nitric Acid 100
$279.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 4, 5000 ppm Ca/K/Mg/Na; 500 mL EW-85012-14 Mfr # RV010706-500N
COMPARE
Ca 5000 ppm, K 5000 ppm, Mg 5000 ppm, and Na 5000 ppm 5% Nitric Acid 500
$546.00USD / Each
Ricca Chemical Company VeriSpec™ ICP Calibration Standard 4; 100 mL EW-85012-18 Mfr # RV010711-100N
COMPARE
Ba 1000 ppm, Ca 1000 ppm, Mg 1000 ppm, and Sr 1000 ppm 5% Nitric Acid 100
$199.00USD / Each
Ricca Chemical Company VeriSpec™ Instrument Calibration Standard 4; 100 mL EW-85012-82 Mfr # RV010775-100N
COMPARE
Ca 5000 ppm, K 5000 ppm, Mg 5000 ppm, and Na 5000 ppm 5% Nitric Acid 100
$204.00USD / Each
Ricca Chemical Company VeriSpec™ Instrument Calibration Standard 4; 500 mL EW-85012-83 Mfr # RV010775-500N
COMPARE
Ca 5000 ppm, K 5000 ppm, Mg 5000 ppm, and Na 5000 ppm 5% Nitric Acid 500
$416.00USD / Each
Ricca Chemical Company VeriSpec™ Instrument Check Standard 4; 100 mL EW-85012-91 Mfr # RV010784-100N
COMPARE
Mo 10 ppm, Sn 10 ppm, Sr 10 ppm, and Ti 10 ppm 2% Nitric Acid 100
$177.00USD / Each
Ricca Chemical Company VeriSpec™ Primary Interference Analytes; 100 mL EW-85012-95 Mfr # RV010788-100N
COMPARE
Al 5000 ppm, Ca 5000 ppm, Fe 2000 ppm, and Mg 5000 ppm 5% Nitric Acid 100
$331.00USD / Each
Ricca Chemical Company VeriSpec™ Primary Interference Analytes; 500 mL EW-85012-96 Mfr # RV010788-500N
COMPARE
Al 5000 ppm, Ca 5000 ppm, Fe 2000 ppm, and Mg 5000 ppm 5% Nitric Acid 500
$598.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 4, 1000 ppm Al/Ca/Ge/Mg; 100 mL EW-85013-22 Mfr # RV010815-100N
COMPARE
Al 1000 ppm, Ca 1000 ppm, Fe 1000 ppm, and Mg 1000 ppm 5% Nitric Acid 100
$229.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 4, 1000 ppm Al/Ca/Ge/Mg; 500 mL EW-85013-23 Mfr # RV010815-500N
COMPARE
Al 1000 ppm, Ca 1000 ppm, Fe 1000 ppm, and Mg 1000 ppm 5% Nitric Acid 500
$441.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 4, 100 ppm As/Be/Pb/Cd; 100 mL EW-85013-32 Mfr # RV010825-100N
COMPARE
As 100 ppm, Be 100 ppm, Cd 100 ppm, and Pb 100 ppm 5% Nitric Acid 100
$173.00USD / Each
Ricca Chemical Company VeriSpec™ Secondary Drnking Water Metals; 100 mL EW-85013-57 Mfr # RV010851-100N
COMPARE
Cu 100 ppm, Fe 30 ppm, Mn 5 ppm, and Zn 500 ppm 2% Nitric Acid and Hydrofluoric Acid 100
$179.00USD / Each
Ricca Chemical Company VeriSpec™ Alternate Metals II; 100 mL EW-85013-62 Mfr # RV010856-100N
COMPARE
Ca 500 ppm, K 100 ppm, Mg 100 ppm, and Na 500 ppm 2% Nitric Acid 100
$172.00USD / Each
Ricca Chemical Company VeriSpec™ Alternate Metals II; 500 mL EW-85013-63 Mfr # RV010857-500N
COMPARE
Ca 500 ppm, K 100 ppm, Mg 100 ppm, and Na 500 ppm 2% Nitric Acid 500
$340.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Environmental Standard 4; 100 mL EW-85014-02 Mfr # RV010899-100N
COMPARE
Ca 1000 ppm, K 1000 ppm, Mg 1000 ppm, and Na 1000 ppm 5% Nitric Acid 100
$184.00USD / Each
Showing 1- 17 of 17