Ricca Chemical Company VeriSpec™ 6-Element Calibration Standard
Multi-element/ion calibration solutions
  • Made to Guide 34 Standards!
$124.00 - $662.00USD / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ ICP Internal Standard 6; 100 mL EW-85011-81 Mfr # RV010674-100N
COMPARE
Bi 100 ppm, In 100 ppm, ⁶Li 100 ppm, Sc 100 ppm, Tb 100 ppm, and Y 100 ppm 5% Nitric Acid 100
$323.00USD / Each
Ricca Chemical Company VeriSpec™ Mixed Cation Standard 6, 1000 ppm Ca; 100 mL EW-85011-96 Mfr # RV010689-100N
COMPARE
Ca²⁺ 1000 ppm, K⁺ 200 ppm, Li⁺ 50 ppm, Mg²⁺ 200 ppm, Na⁺ 200 ppm, and NH₄⁺ 400 ppm Water 100
$217.00USD / Each
Ricca Chemical Company VeriSpec™ Mixed Cation Standard 6, 1000 ppm Ca; 50 mL EW-85011-97 Mfr # RV010689-50N
COMPARE
Ca²⁺ 1000 ppm, K⁺ 200 ppm, Li⁺ 50 ppm, Mg²⁺ 200 ppm, Na⁺ 200 ppm, and NH₄⁺ 400 ppm Water 50
$124.00USD / Each
Ricca Chemical Company VeriSpec™ Mixed Cation Standard 6, 500 ppm Ca/K; 100 mL EW-85011-98 Mfr # RV010691-100N
COMPARE
Ca²⁺ 500 ppm, K⁺ 500 ppm, Li⁺ 50 ppm, Mg²⁺ 250 ppm, Na⁺ 200 ppm, and NH₄⁺ 250 ppm Water 100
$217.00USD / Each
Ricca Chemical Company VeriSpec™ Mixed Cation Standard 6, 10 ppm Ca/K/Mg; 100 mL EW-85012-64 Mfr # RV010757-100N
COMPARE
Ca²⁺ 10 ppm, K⁺ 10 ppm, Li⁺ 1 ppm, Mg²⁺ 10 ppm, Na⁺ 5 ppm, and NH₄⁺ 5 ppm Water 100
$217.00USD / Each
Ricca Chemical Company VeriSpec™ Mixed Cation Standard 6, 100 ppm Ca/K/Li/Na/NH₄/Mg; 100 mL EW-85012-65 Mfr # RV010758-100N
COMPARE
Ca²⁺ 100 ppm, K⁺ 100 ppm, Li⁺ 100 ppm, Mg²⁺ 100 ppm, Na⁺ 100 ppm, and NH₄⁺ 100 ppm Water 100
$217.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 6, Be/Cd/Mn/Pb/Se/Zn; 100 mL EW-85012-72 Mfr # RV010765-100N
COMPARE
Be 50 ppm, Cd 150 ppm, Mn 100 ppm, Pb 500 ppm, Se 200 ppm, and Zn 150 ppm 2% Nitric Acid 100
$221.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 6, Al/Ca/Cr/K/Na/Ni; 100 mL EW-85012-75 Mfr # RV010768-100N
COMPARE
Al 200 ppm, Ca 1000 ppm, Cr 20 ppm, K 400 ppm, Na 200 ppm, and Ni 20 ppm 5% Nitric Acid 100
$214.00USD / Each
Ricca Chemical Company VeriSpec™ Interference Check, Alternate Analytes A; 100 mL EW-85012-97 Mfr # RV010790-100N
COMPARE
Cr 1000 ppm, Cu 1000 ppm, Mn 1000 ppm, Ni 1000 ppm, Ti 1000 ppm, and V 1000 ppm 5% Nitric Acid 100
$382.00USD / Each
Ricca Chemical Company VeriSpec™ Interference Check, Alternate Analytes A; 500 mL EW-85012-98 Mfr # RV010790-500N
COMPARE
Cr 1000 ppm, Cu 1000 ppm, Mn 1000 ppm, Ni 1000 ppm, Ti 1000 ppm, and V 1000 ppm 5% Nitric Acid 500
$662.00USD / Each
Ricca Chemical Company VeriSpec™ Multi-Element Standard 6, Cd/Cr/Cu/Ni/Pb/Zn; 100 mL EW-85013-35 Mfr # RV010828-100N
COMPARE
Cd 10 ppm, Cr 900 ppm, Cu 800 ppm, Ni 200 ppm, Pb 900 ppm, and Zn 2500 ppm 5% Nitric Acid 100
$258.00USD / Each
Ricca Chemical Company VeriSpec™ Elemental Impurities Standard (USP 232) in 5% HCl; 100 mL EW-85013-55 Mfr # RV010849-100N
COMPARE
Ir 100 ppm, Os 100 ppm, Pd 100 ppm, Pt 100 ppm, Rh 100 ppm, and Ru 100 ppm 5% Hydrochloric Acid 100
$384.00USD / Each
Ricca Chemical Company VeriSpec™ Trace Metals Standard III; 100 mL EW-85013-60 Mfr # RV010854-100N
COMPARE
Ba 500 ppm, Ca 500 ppm, K 100 ppm, Mg 100 ppm, Mo 500 ppm, and Na 500 ppm 2% Nitric Acid 100
$202.00USD / Each
Ricca Chemical Company VeriSpec™ Internal Standard 6; 100 mL EW-85013-99 Mfr # RV010896-100N
COMPARE
Ga 20 ppm, In 10 ppm, Ir 10 ppm, Rh 10 ppm, Sc 200 ppm, and Tm 10 ppm 2% Nitric Acid 100
$258.00USD / Each
Showing 1- 14 of 14