Ricca Chemical Company VeriSpec™ Dysprosium ICP Standard
Single-element calibration standard
  • Made to Guide 34 Standards!
$51.50 - $439.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP 1000 ppm in 2% HNO₃; 100 mL EW-85003-43 Mfr # RV010276-100N
COMPARE
1000 ppm 2% Nitric Acid 100
$67.50USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP 1000 ppm in 2% HNO₃; 250 mL EW-85003-44 Mfr # RV010276-250N
COMPARE
1000 ppm 2% Nitric Acid 250
$122.00USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP 1000 ppm in 2% HNO₃; 500 mL EW-85003-45 Mfr # RV010276-500N
COMPARE
1000 ppm 2% Nitric Acid 500
$173.00USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP 1000 ppm in 2% HNO₃; 50 mL EW-85003-46 Mfr # RV010276-50N
COMPARE
1000 ppm 2% Nitric Acid 50
$51.50USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP 10000 ppm in 5% HNO₃; 100 mL EW-85007-13 Mfr # RV010378-100N
COMPARE
10,000 ppm 5% Nitric Acid 100
$136.00USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP 10000 ppm in 5% HNO₃; 250 mL EW-85007-14 Mfr # RV010378-250N
COMPARE
10,000 ppm 5% Nitric Acid 250
$272.00USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP 10000 ppm in 5% HNO₃; 500 mL EW-85007-15 Mfr # RV010378-500N
COMPARE
10,000 ppm 5% Nitric Acid 500
$439.00USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP/MS 100 ppm in 2% HNO₃; 100 mL EW-85010-04 Mfr # RV010517-100N
COMPARE
100 ppm 2% Nitric Acid 100
$83.50USD / Each
Ricca Chemical Company VeriSpec™ Dysprosium Standard for ICP/MS 10 ppm in 2% HNO₃; 100 mL EW-85010-76 Mfr # RV010585-100N
COMPARE
10 ppm 2% Nitric Acid 100
$73.00USD / Each
Showing 1- 9 of 9