Ricca Chemical Company VeriSpec™ Potassium AAS Standard
Single-element calibration standard
  • Made to Guide 34 Standards!
$36.50 - $292.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ Potassium Ionization Buffer for AAS 2% K in 1% HNO₃; 100 mL EW-85000-35 Mfr # RV010010-100N
COMPARE
2% K 1% Nitric Acid 100
$292.00USD / Each
Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 1000 ppm in 2% HCl; 100 mL EW-85000-83 Mfr # RV010028-100N
COMPARE
1000 ppm 2% Hydrochloric Acid 100
$36.50USD / Each
Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 1000 ppm in 2% HCl; 500 mL EW-85000-84 Mfr # RV010028-500N
COMPARE
1000 ppm 2% Hydrochloric Acid 500
$64.00USD / Each
Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 1000 ppm in 2% HNO₃; 100 mL EW-85001-86 Mfr # RV010075-100N
COMPARE
1000 ppm 2% Nitric Acid 100
$36.50USD / Each
Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 1000 ppm in 2% HNO₃; 500 mL EW-85001-87 Mfr # RV010075-500N
COMPARE
1000 ppm 2% Nitric Acid 500
$64.00USD / Each
Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 1000 ppm in 12% Ethanol; 100 mL EW-85002-29 Mfr # RV010119-100N
COMPARE
1000 ppm 12% Ethanol 100
$73.00USD / Each
Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 1000 ppm in 12% Ethanol; 500 mL EW-85002-30 Mfr # RV010119-500N
COMPARE
1000 ppm 12% Ethanol 500
$111.00USD / Each
Ricca Chemical Company VeriSpec™ Potassium Standard for AAS 5 ppb in 1% HNO₃; 50 mL EW-85002-73 Mfr # RV010250-50N
COMPARE
5 ppb 1% Nitric Acid 50
$138.00USD / Each
Showing 1- 8 of 8