Ricca Chemical Company VeriSpec™ Specialty ICP Standards
Multi-element instrument calibration standards
  • Made to Guide 34 Standards!
$64.50 - $441.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Concentration Medium Volume (mL)
Availability Pricing
Ricca Chemical Company VeriSpec™ ICP-MS Internal Standard 8; 100 mL EW-85011-67 Mfr # RV010660-100N
COMPARE
Bi 10 ppm, Ge 10 ppm, In 10 ppm, ⁶Li 10 ppm, Lu 10 ppm, Rh 10 ppm, Sc 10 ppm, and Tb 10 ppm 10% Nitric Acid and Hydrochloric Acid 100
$226.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion ICP-MS Calibration Standard for Set Up 8; 100 mL EW-85013-84 Mfr # RV010880-100N
COMPARE
Be 0.001 ppm, Ce 0.001 ppm, Fe 0.001 ppm, In 0.001 ppm, Li 0.001 ppm, Mg 0.001 ppm, Pb 0.001 ppm, and U 0.001 ppm 2% Nitric Acid and 5% Hydrochloric Acid 100
$212.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion ICP-MS Calibration Standard for Set Up 8; 500 mL EW-85013-85 Mfr # RV010880-500N
COMPARE
Be 0.001 ppm, Ce 0.001 ppm, Fe 0.001 ppm, In 0.001 ppm, Li 0.001 ppm, Mg 0.001 ppm, Pb 0.001 ppm, and U 0.001 ppm 2% Nitric Acid and 5% Hydrochloric Acid 500
$441.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion KED ICP-MS Calibration Standard for Set Up 2; 100 mL EW-85013-86 Mfr # RV010882-100N
COMPARE
Ce 1 ppb and Co 10 ppb 1% Hydrochloric Acid 100
$133.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion KED ICP-MS Calibration Standard for Set Up 2; 500 mL EW-85013-87 Mfr # RV010882-500N
COMPARE
Ce 1 ppb and Co 10 ppb 1% Hydrochloric Acid 500
$263.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion DRC ICP-MS Detection Limit Standard, Blank Solution; 100 mL EW-85013-90 Mfr # RV010886-100N
COMPARE
Blank 0.5% Nitric Acid 100
$64.50USD / Each
Ricca Chemical Company VeriSpec™ Nexion DRC ICP-MS Detection Limit Standard; 100 mL EW-85013-91 Mfr # RV010887-100N
COMPARE
Be 0.001 ppm, Ca 0.001 ppm, Co 0.001 ppm, Fe 0.001 ppm, In 0.001 ppm, and U 0.001 ppm 0.5% Nitric Acid 100
$187.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion KED ICP-MS Detection Limit Standard; 100 mL EW-85013-92 Mfr # RV010888-100N
COMPARE
As 0.01 ppm, Se 0.01 ppm, and V 0.01 ppm 1% Hydrochloric Acid 100
$143.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion AFT ICP-MS Single Element Standard, 2 ppb Fe; 100 mL EW-85013-94 Mfr # RV010890-100N
COMPARE
2 ppb 2% Nitric Acid 100
$104.00USD / Each
Ricca Chemical Company VeriSpec™ Nexion AFT ICP-MS Multi-Element Standard; 100 mL EW-85013-95 Mfr # RV010891-100N
COMPARE
Ag 0.002 ppm, Al 0.002 ppm, As 0.002 ppm, Ba 0.002 ppm, Be 0.002 ppm, Bi 0.002 ppm, Ca 0.002 ppm, Cd 0.002 ppm, Co 0.002 ppm, Cr 0.002 ppm, Cs 0.002 ppm, Cu 0.002 ppm, Fe 0.002 ppm, Ga 0.002 ppm, In 0.002 ppm, K 0.002 ppm, Li 0.002 ppm, Mg 0.002 ppm, Mn 0.002 ppm, Na 0.002 ppm, Ni 0.002 ppm, Pb 0.002 ppm, Rb 0.002 ppm, Se 0.002 ppm, Sr 0.002 ppm, Tl 0.002 ppm, U 0.002 ppm, V 0.002 ppm, and Zn 0.002 ppm 2% Nitric Acid 100
$292.00USD / Each
Showing 1- 10 of 10