Ricca Chemical Methylene Blue, 0.1% (w/v) Aqueous Solution

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$23.60 - $67.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL)
Availability Pricing
Ricca Chemical Company 4841-16 Methylene Blue, 0.1% (w/v) Aqueous Solution (500 mL) EW-86973-46 Mfr # 4841-16
COMPARE
Includes 7220-79-3 and 7732-18-5 500
$45.20USD / Each
Ricca Chemical Company 4841-32 Methylene Blue, 0.1% (w/v) Aqueous Solution (1 L) EW-86980-68 Mfr # 4841-32
COMPARE
Includes 7220-79-3 and 7732-18-5 1000
$67.00USD / Each
Ricca Chemical Company 4841-4 Methylene Blue, 0.1% (w/v) Aqueous Solution (120 mL) EW-86991-10 Mfr # 4841-4
COMPARE
Includes 7220-79-3 and 7732-18-5 120
$23.60USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat