Ricca Chemical ORP Standard +200 mV vs. Ag/AgCl—4M KCl with AgCl filling solution
Fast, simple calibration
  • Use as relative mV standard and calibrate offset of sample
This ORP standard features +200 mV vs. Ag/AgCl with 4M KCl saturated with AgCl filling solution.
$35.70 - $61.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Ricca Chemical Company 5464.5-16 ORP Standard, +200 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (500 mL)
Always in Stock
EW-86974-09 Mfr # 5464.5-16
COMPARE
Includes 7447-40-7, 13746-66-2, 14459-95-1 and 7732-18-5 500 ORP Standard, +200 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (500 mL)
$61.00USD / Each
Ricca Chemical Company 5464.5-4 ORP Standard, +200 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (120 mL) EW-86991-59 Mfr # 5464.5-4
COMPARE
Includes 7447-40-7, 13746-66-2, 14459-95-1 and 7732-18-5 120 ORP Standard, +200 mV vs. Ag/AgCl (4M KCl sat. with AgCl filling solution) (120 mL)
$35.70USD / Each