Ricca Chemical Potassium Chloride Reference Solution C, 1408.8 µS/cm (1408.8 µmho/cm) at 25°C, 0.01 Molar
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$42.90 - $87.40USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL)
Availability Pricing
Ricca Chemical Company 5895-16 Potassium Chloride Reference Solution C, 1408.8 µS/cm at 25°C, (500 mL) EW-86974-78 Mfr # 5895-16
COMPARE
Includes 7447-40-7 and 7732-18-5 500
$42.90USD / Each
Ricca Chemical Company 5895-32 Potassium Chloride Reference Solution C, 1408.8 µS/cm 0.01 Molar (1 L) EW-86981-47 Mfr # 5895-32
COMPARE
Includes 7447-40-7 and 7732-18-5 1000
$87.40USD / Each