Ricca Chemical Potassium Fluoride, 40% (w/v) Aqueous Solution

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$132.25 - $242.75USD / Each

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Ricca Chemical Company 6100-16 Potassium Fluoride, 40% (w/v) Aqueous Solution (500 mL) EW-86974-97 Mfr # 6100-16
COMPARE
Includes 7789-23-3 and 7732-18-5 500 Potassium Fluoride, 40% (w/v) Aqueous Solution (500 mL)
$132.25USD / Each
Ricca Chemical Company 6100-32 Potassium Fluoride, 40% (w/v) Aqueous Solution (1 L) EW-86981-61 Mfr # 6100-32
COMPARE
Includes 7789-23-3 and 7732-18-5 1000 Potassium Fluoride, 40% (w/v) Aqueous Solution (1 L)
$242.75USD / Each
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat