EW-86976-61 Mfr # 8146-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 0.7% (v/v) Aqueous Solution (500 mL)
8697661 EW-86976-62 Mfr # 8149-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 5% (v/v) Aqueous Solution (1 + 19) (500 mL)
8697662 EW-86976-63 Mfr # 8150-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 10% (v/v) Aqueous Solution (1 + 9) (500 mL)
8697663 EW-86976-65 Mfr # 8162-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 16.7% (v/v) Aqueous Solution (1 + 5) (500 mL)
8697665 EW-86976-67 Mfr # 8166-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 20% (w/w) Aqueous Solution (500 mL)
8697667 EW-86976-68 Mfr # 8170-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 25% (v/v) Aqueous Solution (1 + 3) (500 mL)
8697668 EW-86976-69 Mfr # 8180-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (500 mL)
8697669 EW-86976-71 Mfr # 8190-16
COMPARE Includes 7664-93-9 and 7732-18-5 500 Sulfuric Acid, 80% (v/v) Aqueous Solution (4 + 1) (500 mL)
8697671 EW-86983-49 Mfr # 8149-32
COMPARE Includes 7664-93-9 and 7732-18-5 1000 Sulfuric Acid, 5% (v/v) Aqueous Solution (1 + 19) (1 L)
8698349 EW-86983-50 Mfr # 8150-32
COMPARE Includes 7664-93-9 and 7732-18-5 1000 Sulfuric Acid, 10% (v/v) Aqueous Solution (1 + 9) (1 L)
8698350 EW-86983-53 Mfr # 8165-32
COMPARE Includes 7664-93-9 and 7732-18-5 1000 Sulfuric Acid, 20% (v/v) Aqueous Solution (1 + 4), Reagent Grade (1 L)
8698353 EW-86983-54 Mfr # 8170-32
COMPARE Includes 7664-93-9 and 7732-18-5 1000 Sulfuric Acid, 25% (v/v) Aqueous Solution (1 + 3) (1 L)
8698354 EW-86983-55 Mfr # 8180-32
COMPARE Includes 7664-93-9 and 7732-18-5 1000 Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (1 L)
8698355 EW-86984-46 Mfr # 8180-5PT
COMPARE Includes 7664-93-9 and 7732-18-5 2500 Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (2.5 L)
$203.50 USD / Each
8698446 8698446 EW-86988-20 Mfr # 8150-1
COMPARE Includes 7664-93-9 and 7732-18-5 4000 Sulfuric Acid, 10% (v/v) Aqueous Solution (1 + 9) (1 gal)
$126.75 USD / Each
8698820 8698820 EW-86988-22 Mfr # 8170-1
COMPARE Includes 7664-93-9 and 7732-18-5 4000 Sulfuric Acid, 25% (v/v) Aqueous Solution (1 + 3) (1 gal)
$176.75 USD / Each
8698822 8698822 EW-86988-23 Mfr # 8180-1
COMPARE Includes 7664-93-9 and 7732-18-5 4000 Sulfuric Acid, 50% (v/v) Aqueous Solution (1 + 1) (1 gal)
$235.00 USD / Each
8698823 8698823