Positive-Displacement Roller Pumps
Ideal for high temperature applications where some particulates are present
  • NSF approved models are ideal for food applications
  • No metal to metal contact means less wear and a longer service life
  • Pumps are self-priming — pumps instantly with a dry suction lift of up to 10 feet
Economical yet highly versatile pumps are ideal for fluid spraying and transfer applications. NSF models feature mechanical seals for long life. Pumps are specifically designed to prevent clogging. Ideal for pumping hot oils and low shear oils. Pumps do not require foot valves.
Ni-resist is an alloy that contains 13% nickel for added corrosion resistance. Order a metallic pump below for this feature.
$810.75 - $1,126.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Air Inlet Pressure (PSI) Max Flow Rate (GPM) Max Motor RPM (rpm)
Availability Pricing
COMPARE
3.8 1725
$889.50USD / Each
COMPARE
5.8 1725
$896.50USD / Each
COMPARE
8 1725
$1,126.50USD / Each
COMPARE
7.5 1900
$810.75USD / Each