Saccharin Sodium

  • Assay - 98.0 to 101.0%
  • Alkalinity - To pass test
  • Benzoate and Salicylate - To pass test
  • Readily Carbonizable Substances - To pass test
  • Heavy Metals (as Pb) - ≤0.001%
  • Selenium - ≤0.003%
  • Toluene Sulfonamides - <0.0025%
  • Water - ≤15.0%
  • Organic Volatile Impurities - Meets the requirements
$342.00USD / Each

1 variations of this product are available.

See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical S394500 Saccharin Sodium (Powder/USP/FCC) (500g) EW-88081-41 Mfr # S394500
COMPARE
6455-57-3 FCC or USP
$342.00USD / Each
Showing 1- 1 of 1

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat