This product is now discontinued, View Related Products

Safety Light Curtains

Provide protection against category 4 hazards
  • Meets ANSI/RIA, OSHA, IEC, EN, UL, and CSA standards
  • Units are self-contained – no control box required
  • Optional setting console makes setup/operation simple
Use these independently certified light curtains for protection against life-threatening category 4 hazards. These light curtains are available in incremental sizes, by number of beams, so you can specify the exact protective height you need saving on each system. Indicator LED’s show status, error, and light intensity on both the emitter and the receiver. The slim profile is space efficient and ideal for applications requiring category 4 hazard protection. Series connectable models are available upon request, allowing for up to three sets of 240 beams to be connected in series, and provide an auto and manual reset function.
Use your own, or order the optional safety relay, and cables, (below) to complete your light curtain system, and reduce wiring time and eliminate possible improper wiring. The optional setting console may be added to provide setup and operation flexibility. The fixed blanking of one to nine beams, one to three zones, and a teach function offer design flexibility that will meet your installation needs. Floating blanking of one to three beams, and the teach function provide a sequential mode ensuring that beams have to be broken consecutively (floating and fixed blanking requires optional setting console). After powering the safety light curtain system on the diagnostic function feature performs a complete self-test of the light curtain system within one second, and performs a self-test periodically during operation.
What’s included: One each emitter and receiver light curtain devices.
Call For Price

12 variations of this product are available.

See All Product Options Below
12 PRODUCT OPTIONS
Compareitem Availability Pricing
Omron F3SN-A0217P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-00 Mfr # F3SN-A0217P25
COMPARE
Call For Price
Omron F3SN-A0307P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-02 Mfr # F3SN-A0307P25
COMPARE
Call For Price
Omron F3SN-A0457P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-04 Mfr # F3SN-A0457P25
COMPARE
Call For Price
Omron F3SN-A0607P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-06 Mfr # F3SN-A0607P25
COMPARE
Call For Price
Omron F3SN-A0757P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-08 Mfr # F3SN-A0757P25
COMPARE
Call For Price
Omron F3SN-A0907P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-10 Mfr # F3SN-A0907P25
COMPARE
Call For Price
Omron F3SN-A1057P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-12 Mfr # F3SN-A1057P25
COMPARE
Call For Price
Omron F3SN-A1207P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-14 Mfr # F3SN-A1207P25
COMPARE
Call For Price
Omron F3SN-A1357P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-16 Mfr # F3SN-A1357P25
COMPARE
Call For Price
Omron F3SN-A1507P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-18 Mfr # F3SN-A1507P25
COMPARE
Call For Price
Omron F3SN-A1657P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-20 Mfr # F3SN-A1657P25
COMPARE
Call For Price
Omron F3SN-A1807P25 Safety Light Curtain, 13 beam - 217 mm Protective height EW-49801-22 Mfr # F3SN-A1807P25
COMPARE
Call For Price
Showing 1- 12 of 12

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat