Saint Gobain® Tygon® A-60-F IB Pressure Transfer Tubing
Flexibility at extreme temperatures
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
Ideal for chemical transfer, pesticide/fertilizer delivery, water feed lines, food/dairy processing, and water purification lines. The tube features excellent resistance to acidic and alkali substances, ozone, UV light, and abrasive media. It is nonconductive, heat-sealable, non-aging, and nonoxidizing. In addition, the tube maintains flexibility even at extreme temperatures.
Temperature range is -60 to 275°F (-51 to 135°C).
Can be sterilized with autoclave.
$378.00 - $1,396.00USD / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Saint Gobain Transfer Tubing, Tygon® A-60-F IB, 3/8" ID x 5/8" OD; 50 Ft
Buy More Save More
EW-95715-01 Mfr # APW00029
COMPARE
105 3/8 5/8
$467.00USD / Pkg of 1
Saint Gobain Transfer Tubing, Tygon® A-60-F IB, 1/2" ID x 3/4" OD; 50 Ft
Buy More Save More
EW-95715-02 Mfr # APW00038
COMPARE
100 1/2 3/4
$651.00USD / Pkg of 1
Saint Gobain Transfer Tubing, Tygon® A-60-F IB, 1" ID x 1-3/8" OD; 50 Ft
Buy More Save More
EW-95715-05 Mfr # APW00064
COMPARE
75 1 1
$1,396.00USD / Pkg of 1
Saint Gobain Transfer Tubing, Tygon® A-60-F IB, 1/4" ID x 1/2" OD; 50 Ft EW-95715-06 Mfr # APW00019
COMPARE
125 1/4 1/2
$378.00USD / Pkg of 1