Salt Brine Plain Form Hydrometers
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
ITEM(S) ON SALE
$30.34 - $52.87USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Salt Brine Hydrometer; 0/100 Percent by Saturation
$30.34USD / Each
$35.70USD / Each
COMPARE
Salt Brine Hydrometer; 0/26.5 Percent by Weight
$52.87USD / Each
$62.20USD / Each