BriskHeat® Samox High-Temperature Heating Tapes leads opposite ends-120 VAC
Flexible, multipurpose, direct-contact heaters are ideal for use on conductive surfaces. The 120 VAC tapes include a 120 VAC two-prong plug. Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
  • Maximum exposure temperature of 1400°F (760°C)
  • Choose from power leads on same end or on opposite ends
  • Heavy-duty insulated design
  • Nongrounded
Resistance wires are double insulated with braided SAMOX yarns, then knit into flat tapes. SAMOX fabric also covers tapes to provide extra protection against abrasion. Tapes include ties at each end.
These high-temperature heating tapes are made of resistance wires that are insulated with braided fiberglass knitted into a flat tape and covered with heavy fiberglass fabric.
The 240 VAC tapes with an opposite-end lead configuration are supplied with bare leads.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately.
$181.00 - $396.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat 051100 Samox Heating Tape, Maximum Exposure Temperature1400 and Lt; Deg and Gt; F (760 and Lt; Deg and G EW-03106-07 Mfr # TBWH-051-100
COMPARE
10 1400 120
$298.00USD / Each
BriskHeat 101060L Samox Heating Tape 1400F (760C), 1" Wide By 6ft Length, Leads Opposite Ends EW-03106-12 Mfr # TBWH-101-060
COMPARE
6 1400 120
$291.00USD / Each
BriskHeat TBWH051-020 Samox Insulated Heating Tape Leads On Opposite Ends, 2' x 1/2" , 156 Watts, 120 V EW-03115-20 Mfr # TBWH051-020
COMPARE
2 1400 120
$193.00USD / Each
BriskHeat TBWH051-040 Samox Insulated Heating Tape Leads On Opposite Ends, 4' x 1/2" , 312 Watts, 120 V EW-03115-40 Mfr # TBWH051-040
COMPARE
4 1400 120
$215.00USD / Each
BriskHeat TBWH051-060 Samox Insulated Heating Tape Leads On Opposite Ends, 6' x 1/2" , 468 Watts, 120 V EW-03115-60 Mfr # TBWH051-060
COMPARE
6 1400 120
$258.00USD / Each
BriskHeat TBWH051-080 Samox Insulated Heating Tape Leads On Opposite Ends, 8' x 1/2" , 624 Watts, 120 V EW-03115-80 Mfr # TBWH051-080
COMPARE
8 1400 120
$274.00USD / Each
BriskHeat TBWH101-020 Samox Insulated Heating Tape Leads On Opposite Ends, 2' x 1" , 314 Watts, 120 V EW-03117-20 Mfr # TBWH101-020
COMPARE
2 1400 120
$181.00USD / Each
BriskHeat TBWH101-040 Samox Insulated Heating Tape Leads On Opposite Ends, 4' x 1" , 628 Watts, 120 V EW-03117-40 Mfr # TBWH101-040
COMPARE
4 1400 120
$209.00USD / Each
BriskHeat TBWH171-040 Samox Insulated Heating Tape Leads On Opposite Ends, 4' x 1-3/4" , 628 Watts, 120 V EW-03118-40 Mfr # TBWH171-040
COMPARE
4 1400 120
$396.00USD / Each
Showing 1- 9 of 9