Sandpiper Air-Powered Double-Diaphragm Polypropylene Pumps
Move fluids with viscosities up to 11,000 cp
  • Air-powered motor eliminates sparking concerns associate with electrical pumps
  • Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing
  • Non-metallic pumps are ideally suited for highly corrosive fluids intermittent/on-demand, portable low abrasive fluids, and suspended solids
  • Safely handles abrasive fluids and slurries — ideal for high-viscosity and shear sensitive materials
  • Encapsulated muffler provides excellent sound dampening
These pumps can move high-viscosity fluids such as paint, sludge, and sewage. However, flow rate and pressure depends on the application. If you have questions about the compatibility of your high-viscosity fluid, call our Application Specialists for more information.
Order air filters/regulators to remove impurities from your air line and to help regulate working pressure. Combination air filter/regulators feature polypropylene filter, 0- to 160-psi gauge, and a polycarbonate bowl. Temperature range is 0 to 125°F (17 to 51°C).
$459.70 - $1,356.50USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Wetted Materials
Availability Pricing
Warren Rupp Air-Powered Double-Diaphragm PP Pump, Santoprene; 14 GPM EW-74000-00 Mfr # S05B2P1TPNS000
COMPARE
14 Polypropylene, Santoprene®
$915.25USD / Each
Warren Rupp Air-Powered Double-Diaphragm PP Pump, Santoprene; 23 GPM EW-74000-11 Mfr # S07B1P1PPNS000.
COMPARE
23 Polypropylene, Santoprene®
$1,041.50USD / Each
Warren Rupp S07B1P2PPNS000. Air-Powered Double-Diaphragm PP Pump, PTFE; 23 GPM EW-74000-13 Mfr # S07B1P2PPNS000.
COMPARE
23 Polypropylene, PTFE
$1,314.00USD / Each
Warren Rupp SO5B2P2TPNS000 Air-Powered Double-Diaphragm PP Pump, PTFE; 14 GPM EW-74000-42 Mfr # SO5B2P2TPNS000
COMPARE
14 Polypropylene, PTFE
$1,356.50USD / Each
Warren Rupp PB.25,TS3PP. Air-Powered Double-Diaphragm PP Pump, Santoprene; 4 GPM
Always in Stock
EW-74000-70 Mfr # PB 1/4 TS4PP.
COMPARE
4 Polypropylene, Santoprene®
$459.70USD / Each
Warren Rupp PB.25,TT3PP. Air-Powered Double-Diaphragm PP Pump, PTFE; 4 GPM
Always in Stock
EW-74000-72 Mfr # PB 1/4 TT4PP
COMPARE
4 Polypropylene, PTFE
$600.25USD / Each