Sani-Lock 316L Stainless Steel Quick-Connect Clamps
No hinge clamp required – rapid one-hand connection
  • Snap-in-place locking arms secure fitting, while safety pins prevent accidental release
  • Fully compatible with sanitary Tri-Clamp® fittings
  • 150 psi (10.3 bar) working line pressure
Converts any connection into a sanitary "quick-connect" connection. Sani-Lock provides the convenience and efficiency of a quick-connect while maintaining the integrity of a sanitary connection. Use with Tuf-Flex® gaskets for guaranteed leak-proof alignment.(sold separately below) Automatic alignment of the Tuf-Flex gasket eliminates over and under compression of gaskets. The locking arms snap-in-place while creating a secure connection.
$385.00 - $646.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Rubberfab Sani-Lock Quick-Connect, 1/2" Sani-Lock x 1/2" Tri-Clamp Adapter EW-31607-70 Mfr # 08TCL-08TC-SS
COMPARE
Sani-Lock Quick-Connect, 1/2" Sani-Lock x 1/2" Tri-Clamp Adapter
$385.00USD / Each
Rubberfab Sani-Lock Quick-Connect, 3/4" Sani-Lock to 3/4" Tri-Clamp Adapter EW-31607-76 Mfr # 12HB-12TC-SS
COMPARE
Sani-Lock Quick-Connect, 3/4" Sani-Lock to 3/4" Tri-Clamp Adapter
$385.00USD / Each
Rubberfab Sani-Lock Quick-Connect, 1" Sani-Lock x 1-1/2" Tri-Clamp Adapter EW-31607-84 Mfr # 16HB-24TC-SS
COMPARE
Sani-Lock Quick-Connect, 1" Sani-Lock x 1-1/2" Tri-Clamp Adapter
$646.00USD / Each