Sanitary Ball Valves with Tri-Clamp Connections
  • Wetted parts: 316 stainless steel with a sanitary finish and PTFE seals.
Ball valves feature a unique design that allows for easy in-line servicing. Valve seats are made of fully encapsulated PTFE. Each valve includes a locking handle.
$160.50 - $459.40USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Clamp Size (in) Cv Max Pressure (PSI)
Availability Pricing
COMPARE
0.75 40 1000
$160.50USD / Each
COMPARE
1 70 1000
$173.60USD / Each
COMPARE
1.5 120 1000
$380.30USD / Each
COMPARE
2 340 1000
$459.40USD / Each