Sanitary Plastic Ball Valves
Precision machined and tested to ensure leak-free connections
  • Standard Tri-Clamp connections are easy to plumb into sanitary fluid paths
  • Smooth flow path minimizes turbulence and pressure loss
  • FDA 21 CFR 177.1520 compliant; NSF listed
  • Valve design permits full flow and pressure in either direction
These industry standard valves feature PTFE bearings and seats to virtually eliminate friction and minimize wear. Fully concentric polished ball further ensures leak-free performance. Available in polyproplyene (PP) or PVDF body material with EPDM seals.
$364.00 - $1,419.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Clamp Size (in) Cv Wetted Materials
Availability Pricing
COMPARE
0.75 20 Polypropylene, EPDM
$364.00USD / Each
COMPARE
1 40 Polypropylene, EPDM
$380.00USD / Each
COMPARE
1 40 Polypropylene, EPDM
$440.00USD / Each
COMPARE
1.5 100 Polypropylene, EPDM
$490.00USD / Each
COMPARE
2 120 Polypropylene, EPDM
$646.00USD / Each
COMPARE
0.75 20 PVDF, EPDM
$706.00USD / Each
COMPARE
1 40 PVDF, EPDM
$732.00USD / Each
COMPARE
1 40 PVDF, EPDM
$884.00USD / Each
COMPARE
1.5 100 PVDF, EPDM
$1,075.00USD / Each
COMPARE
2 120 PVDF, EPDM
$1,419.00USD / Each
Showing 1- 10 of 10