Sartorius® Cubis® II (MCA) Analytical Balances with Software
Long-term flexibility for regulatory and laboratory needs
  • Fully customizable hardware, software, and connectivity
  • Complete data integrity for confident compliance
  • Motorized auto-leveling reduces errors
  • Error-free operation by assessing and correcting relevant factors
  • Electronic signature in compliance with 21 CFR Part 11
  • Safe data transfer
  • Advanced MCA user interface features large color TFT touch screen with swipe functionality
Sartorius® Cubis® II MCA analytical balances are completely customizable to align with your unique application and compliance requirements while ensuring reliable results. They offer a max capacity between 120 g and 520 g with a scale interval range of 0.01 mg to 0.1 mg. Innovative technology includes automatic motorized leveling, motion controlled and illuminated draft shield, and climate sensor for environmental monitoring to ensure error-free operation.
Large 7” color touch-screen display allows you to touch, scroll, and swipe with factory-installed weighing applications. The status center shows all information about your balance and environmental conditions as well as detailed help and support in case of warnings and errors.
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Sartorius MCA324S-2S00-U Cubis® II Analytical Balance, 320 g x 0.1 mg; Manual Glass Draft Shield EW-39742-08 Mfr # MCA324S-2S00-U
COMPARE
External 320 0.1
$9,724.00USD / Each
$11,050.00USD / Each
Sartorius MCA124S-2S00-U Cubis® II Analytical Balance, 120 g x 0.1 mg; Manual Glass Draft Shield EW-39742-09 Mfr # MCA124S-2S00-U
COMPARE
External 120 0.1
$7,392.00USD / Each
$8,400.00USD / Each
Sartorius MCA524S-2S00-I Cubis® II Analytical Balance, 520 g x 0.1 mg; Auto Glass Draft Shield, Integrated Ionizer EW-39742-10 Mfr # MCA524S-2S00-I
COMPARE
External 520 0.1
$15,021.60USD / Each
$17,070.00USD / Each
Sartorius MCA324S-2S00-I Cubis® II Analytical Balance, 320 g x 0.1 mg; Auto Glass Draft Shield, Integrated Ionizer EW-39742-11 Mfr # MCA324S-2S00-I
COMPARE
External 320 0.1
$13,613.60USD / Each
$15,470.00USD / Each