This product is now discontinued, View Related Products

Sartorius Entris Toploading Balances

Affordability without compromising reliability and quality
  • Perform basic weighing with below balance, counting, percent, animal weighing, and density determination functions
  • Secure balance to work station with the anti-theft feature
  • Functional in 7 languages
  • Document critical weights via RS-232 communication with PC or printer
  • Cleanup is easy with removable pan and draft shield
  • Backlit high-contrast display delivers excellent visibility in any lighting conditions
Call For Price

24 variations of this product are available.

See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Sartorius ENTRIS153I-1SUS Precision Toploading Balance 150 g x 0.001g, Internal Calibration EW-11952-23 Mfr # ENTRIS153I-1SUS
COMPARE
150 120 / 220 0.001
Call For Price
Sartorius ENTRIS323I-1SUS Precision Toploading Balance 320 g x 0.001g, Internal Calibration EW-11952-24 Mfr # ENTRIS323I-1SUS
COMPARE
320 120 / 220 0.001
Call For Price
Sartorius ENTRIS423I-1SUS Precision Toploading Balance 420 g x 0.001g, Internal Calibration EW-11952-25 Mfr # ENTRIS423I-1SUS
COMPARE
420 120 / 220 0.001
Call For Price
Sartorius ENTRIS623I-1SUS Precision Toploading Balance 620 g x 0.001g, Internal Calibration EW-11952-26 Mfr # ENTRIS623I-1SUS
COMPARE
620 120 / 220 0.001
Call For Price
Sartorius ENTRIS822I-1SUS Toploader Balance 820 g x 0.01g, Internal Calibration EW-11952-27 Mfr # ENTRIS822I-1SUS
COMPARE
820 120 / 220 0.01
Call For Price
Sartorius ENTRIS2202I-1SUS Toploader Balance 2202g x 0.01g, Internal Calibration EW-11952-28 Mfr # ENTRIS2202I-1SUS
COMPARE
2200 120 / 220 0.01
Call For Price
Sartorius ENTRIS4202I-1SUS Toploader Balance 4200 g x 0.01g, Internal Calibration EW-11952-31 Mfr # ENTRIS4202I-1SUS
COMPARE
4200 120 / 220 0.01
Call For Price
Sartorius ENTRIS6202I-1SUS Toploader Balance 6200 g x 0.01g, Internal Calibration EW-11952-32 Mfr # ENTRIS6202I-1SUS
COMPARE
6200 120 / 220 0.01
Call For Price
Sartorius ENTRIS2201I-1SUS Toploader Balance 2200 g x 0.1g, Internal Calibration EW-11952-33 Mfr # ENTRIS2201I-1SUS
COMPARE
2200 120 / 220 0.1
Call For Price
Sartorius ENTRIS5201I-1SUS Toploader Balance 5200 g x 0.1g, Internal Calibration EW-11952-34 Mfr # ENTRIS5201I-1SUS
COMPARE
5200 120 / 220 0.1
Call For Price
Sartorius ENTRIS8201I-1SUS Toploader Balance 8200 g x 0.1g, Internal Calibration EW-11952-35 Mfr # ENTRIS8201I-1SUS
COMPARE
8200 120 / 220 0.1
Call For Price
Sartorius ENTRIS153-1SUS Precision Toploading Balance 150g x 0.001g, Ext Calibration EW-11955-23 Mfr # ENTRIS153-1SUS
COMPARE
150 115 / 230 0.001
Call For Price
Sartorius ENTRIS323-1SUS Precision Toploading Balance 320g x 0.001g, Ext Calibration EW-11955-24 Mfr # ENTRIS323-1SUS
COMPARE
320 115 / 230 0.001
Call For Price
Sartorius ENTRIS423-1SUS Precision Toploading Balance 420g x 0.001g, Ext Calibration EW-11955-25 Mfr # ENTRIS423-1SUS
COMPARE
420 115 / 230 0.001
Call For Price
Sartorius ENTRIS822-1SUS Toploader Balance 820g x 0.01g, Ext Calibration EW-11955-27 Mfr # ENTRIS822-1SUS
COMPARE
820 115 / 230 0.01
Call For Price
Sartorius ENTRIS2202-1SUS Toploader Balance 2202g x 0.01g, Ext Calibration EW-11955-28 Mfr # ENTRIS2202-1SUS
COMPARE
2200 115 / 230 0.01
Call For Price
Sartorius ENTRIS3202-1SUS Toploader Balance 3200g x 0.01g, Ext Calibration 11955-29 EW-11955-29 Mfr # ENTRIS3202-1SUS
COMPARE
3200 115 / 230 0.01
Call For Price
Sartorius ENTRIS4202-1SUS Toploader Balance 4200g x 0.01g, Ext Calibration EW-11955-31 Mfr # ENTRIS4202-1SUS
COMPARE
4200 115 / 230 0.01
Call For Price
Sartorius ENTRIS6202-1SUS Toploader Balance 6200g x 0.01g, Ext Calibration EW-11955-32 Mfr # ENTRIS6202-1SUS
COMPARE
6200 115 / 230 0.01
Call For Price
Sartorius ENTRIS2201-1SUS Toploader Balance 2200g x 0.1g, Ext Calibration EW-11955-33 Mfr # ENTRIS2201-1SUS
COMPARE
2200 115 / 230 0.1
Call For Price
Showing 1 - 20 of 24

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat