Sartorius Quintix Toploading Balances
An outstanding combination of design and technology that provides excellent performance in the laboratory
  • Touchscreen/swipe technology enables users to easily navigate through all balance operations
  • Weighing applications are integrated into the balance menu and available at the touch of a key
  • Simple menu navigation with icon-driven menus
  • Monolithic weigh cell provides superior durability and reliability with precise weighing results
  • ISO/GLP-compliant recording feature complies with documentation requirements
  • Feature isoCAL—a built-in fully automatic internal calibration and balance adjustment function
  • Mini USB interface port enables communication with a PC or printer
ITEM(S) ON SALE
$1,469.60 - $3,722.40USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Sartorius QUINTIX513-1S Toploading Balance, 510g x 0.001ginternal Calibration EW-11800-78 Mfr # QUINTIX513-1S
COMPARE
510 100 to 240 0.001
$3,141.60USD / Each
$3,570.00USD / Each
Sartorius QUINTIX313-1S Toploading Balance, 310g x 0.001ginternal Calibration EW-11800-79 Mfr # QUINTIX313-1S
COMPARE
310 100 to 240 0.001
$2,323.20USD / Each
$2,640.00USD / Each
Sartorius QUINTIX213-1S Toploading Balance, 210g x 0.001ginternal Calibration EW-11800-80 Mfr # QUINTIX213-1S
COMPARE
210 100 to 240 0.001
$2,173.60USD / Each
$2,470.00USD / Each
Sartorius QUINTIX5102-1S Toploading Balance, 5100g x 0.01ginternal Calibration EW-11800-81 Mfr # QUINTIX5102-1S
COMPARE
5100 100 to 240 0.01
$3,185.60USD / Each
$3,620.00USD / Each
Sartorius QUINTIX3102-1S Toploading Balance, 3100g x 0.01ginternal Calibration EW-11800-82 Mfr # QUINTIX3102-1S
COMPARE
3200 100 to 240 0.01
$2,569.60USD / Each
$2,920.00USD / Each
Sartorius QUINTIX2102-1S Toploading Balance, 2100g x 0.01ginternal Calibration EW-11800-83 Mfr # QUINTIX2102-1S
COMPARE
2200 100 to 240 0.01
$2,323.20USD / Each
$2,640.00USD / Each
Sartorius QUINTIX1102-1S Toploading Balance, 1100g x 0.01ginternal Calibration EW-11800-84 Mfr # QUINTIX1102-1S
COMPARE
1102 100 to 240 0.01
$2,076.80USD / Each
$2,360.00USD / Each
Sartorius QUINTIX612-1S Toploading Balance 612, 610g x 0.01ginternal Calibration EW-11800-85 Mfr # QUINTIX612-1S
COMPARE
610 100 to 240 0.01
$1,830.40USD / Each
$2,080.00USD / Each
Sartorius QUINTIX5101-1S Topload Balance 5101, 5100g x 0.1ginternal Calibration EW-11800-86 Mfr # QUINTIX5101-1S
COMPARE
5100 100 to 240 0.1
$1,830.40USD / Each
$2,080.00USD / Each
Sartorius QUINTIX5100-1S Toploading Balance, 5100g x 1.0g, 100-240V EW-11800-87 Mfr # QUINTIX5100-1S
COMPARE
5100 100 to 240 1
$1,469.60USD / Each
$1,670.00USD / Each
Sartorius Quintix613-1S Toploading Balance 610Ginternal Calibration EW-11955-05 Mfr # QUINTIX613-1S
COMPARE
610 115 0.001
$3,308.80USD / Each
$3,760.00USD / Each
Sartorius Quintix412-1S Toploading Balance 410g x 0.01ginternal Calibration EW-11955-06 Mfr # QUINTIX412-1S
COMPARE
410 115 0.01
$1,592.80USD / Each
$1,810.00USD / Each
Sartorius Quintix6102-1S Toploading Balance 6100g x 0.01ginternal Calibration EW-11955-07 Mfr # QUINTIX6102-1S
COMPARE
6100 115 0.01
$3,722.40USD / Each
$4,230.00USD / Each
Sartorius Quintix2101-1S Toploading Blanace 2100g x 0.1ginternal Calibration EW-11955-08 Mfr # QUINTIX2101-1S
COMPARE
2100 115 0.1
$1,592.80USD / Each
$1,810.00USD / Each
Sartorius Quintix6101-1S Toploading Balance 6100g x 0.1ginternal Calibration EW-11955-09 Mfr # QUINTIX6101-1S
COMPARE
6100 115 0.1
$2,200.00USD / Each
$2,500.00USD / Each
Showing 1- 15 of 15