Sartorius Tacta® Multi-Channel Pipettors
Superb comfort with effortless volume adjustment and locking
  • Achieve reliable results with easy calibration adjustment for various fluid types
  • Hold comfortably with the ergonomically designed handle
  • Autoclave fully without disassembly for quick and easy cleaning
This premium mechanical pipette has the Optiload feature in both single- and multichannel models for light tip loading with perfect tip sealing. The Optiject soft-tip ejection feature detaches the tip in a smooth, controlled manner. Optilock’s dual-function volume lock offers protection against accidental volume changes.
Use the easy-to-read, four-digit volume display to set exact volumes. Different volume options are color-coded for quick selection of corresponding pipette tips. Safe-Cone filters (included in models above 10 μL) protect against contamination. Pipettors are made of materials which have high chemical and UV resistance to ensure a long life span.
$1,300.00 - $1,530.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Number Of Channels
Availability Pricing
Sartorius Tacta® LH-729120 Eight-Channel, Adjustable-Volume Pipette; 0.5 to 10 µL EW-24505-41 Mfr # LH-729120
COMPARE
10 0.5 8
$1,300.00USD / Each
Sartorius Tacta® LH-729130 Eight-Channel, Adjustable-Volume Pipette; 5 to 100 µL EW-24505-42 Mfr # LH-729130
COMPARE
100 5 8
$1,300.00USD / Each
Sartorius Tacta® LH-729220 Twelve-Channel, Adjustable-Volume Pipette; 0.5 to 10 µL EW-24505-44 Mfr # LH-729220
COMPARE
10 0.5 12
$1,530.00USD / Each
Sartorius Tacta® LH-729230 Twelve-Channel, Adjustable-Volume Pipette; 5 to 100 µL EW-24505-45 Mfr # LH-729230
COMPARE
100 5 12
$1,530.00USD / Each
Sartorius Tacta® LH-729240 Twelve-Channel, Adjustable-Volume Pipette; 30 to 300 µL EW-24505-46 Mfr # LH-729240
COMPARE
300 30 12
$1,530.00USD / Each